Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04233249_s1
          See other BEND3P3 GE Assays ›
          Gene Symbol:
          BEND3P3
          Gene Name:
          BEN domain containing 3 pseudogene 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 79682975 - 79688897 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04233249_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: BEND3P3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027512.1 1 1354 91

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC105378385 LOC105378385 -50.8kb Anti-sense Chr. 10 - 79628757 - 79663185 on NCBI Build GRCh38
          Downstream NUTM2B antisense RNA 1 NUTM2B-AS1 2.6kb Anti-sense Chr. 10 - 79691500 - 79826594 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1000008210.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC10000571.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 706051 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 656593 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 885351 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 688673 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 706776 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 272906 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 313273 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 8712 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 87161 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 49628 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_5182 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_5181 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3396646 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27728883 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16360651 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P16360651 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P27728883 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR10006171.hg.1 NR_027512.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR10006171.hg.1 TC10000571.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1000008210.hg.1 ENST00000611030.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008210.hg.1 NR_027512.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008210.hg.1 NR_027512.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000007614.hg.1 ENST00000421591.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007614.hg.1 ENST00000421591.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007614.hg.1 ENST00000449695.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007614.hg.1 ENST00000449695.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000007614.hg.1 ENST00000449695.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011184.hg.1 ENST00000452173.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011184.hg.1 ENST00000623504.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000011184.hg.1 ENST00000623504.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0900007948.hg.1 ENST00000414425.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000008210.hg.1 ENST00000611030.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008210.hg.1 NR_027512.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008210.hg.1 NR_027512.1 Refseq
          Affymetrix Human Clariom D Assay TC1000007614.hg.1 ENST00000421591.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000007614.hg.1 ENST00000421591.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000007614.hg.1 ENST00000449695.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000007614.hg.1 ENST00000449695.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000007614.hg.1 ENST00000449695.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000155571.hg.1 NR_027512.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000155571.hg.1 TC1000008210.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000155571.hg.1 NR_027512.1 Refseq
          Affymetrix Human Clariom D Assay TC1000011184.hg.1 ENST00000452173.2 Ensembl
          Affymetrix Human Clariom D Assay TC1000011184.hg.1 ENST00000623504.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000011184.hg.1 ENST00000623504.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000155570.hg.1 ENST00000611030.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000155570.hg.1 NR_027512.1 Refseq
          Affymetrix Human Clariom D Assay PSR1000155570.hg.1 TC1000008210.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1000155570.hg.1 ENST00000611030.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1000155570.hg.1 NR_027512.1 Refseq
          Affymetrix Human Clariom D Assay TC0900007948.hg.1 ENST00000414425.2 Ensembl
          Human Genome U95 Set 79218_at - -
          Human Genome U95 Set 79218_PM_at - -
          Human Genome U95 Set 70728_PM_r_at - -
          Human Genome U95 Set 70728_r_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A044176 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A044176 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0