Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04233300_g1
          See other SCGB1B2P GE Assays ›
          Gene Symbol:
          SCGB1B2P
          Gene Name:
          secretoglobin family 1B member 2, pseudogene
          Gene Aliases:
          SCGB4A1P
          Chromosome Location:
          Chr.19: 34576733 - 34677031 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04233300_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SCGB1B2P
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027620.3 3 - 4 1785 95
          GenBank mRNA CA429783.1 - 1 - 2 95
          CD173361.1 - 2 - 3 375 95
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA HY240171.1 - 1 - 2 Chr. 19 - 34576733 - 34577431 [+] on Build GRCh38 95

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream Wilms tumor 1 interacting protein WTIP -67.6kb Anti-sense Chr. 19 - 34481935 - 34509147 on NCBI Build GRCh38
          Downstream secretoglobin family 2B member 3,
          pseudogene
          SCGB2B3P C2B,
          SCGB4A3P
          68.4kb Anti-sense Chr. 19 - 34645873 - 34646052 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3340868 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3340869 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_5183 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_5184 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 32112 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 18714 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3340868 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3340869 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3340868 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3340869 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945071 - -
          Agilent Whole Chr array_18 19 21 A_17_P17132714 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945055 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945062 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571842 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571837 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945070 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571852 - -
          Agilent Whole Chr array_18 19 21 A_17_P17132691 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571885 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945075 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945050 - -
          Agilent Whole Chr array_18 19 21 A_17_P17132704 - -
          Agilent Whole Chr array_18 19 21 A_17_P17132687 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945052 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945067 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945059 - -
          Agilent Whole Chr array_18 19 21 A_17_P17132695 - -
          Agilent Whole Chr array_18 19 21 A_17_P17132701 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945048 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945047 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945056 - -
          Agilent Whole Chr array_18 19 21 A_17_P17132699 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945053 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571840 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571826 - -
          Agilent Whole Chr array_18 19 21 A_17_P17132688 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945058 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945054 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945046 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945063 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571873 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571876 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945065 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571864 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945074 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571883 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945057 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945076 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945068 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945077 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571884 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571848 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945064 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571844 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945049 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571841 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571882 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945072 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945073 - -
          Agilent Whole Chr array_18 19 21 A_17_P17132683 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571846 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945051 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571838 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945060 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945066 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945045 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945069 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10945054 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10945056 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31571864 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10945067 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31571842 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17132701 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31571876 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31571852 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10945053 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31571846 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10945059 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10945074 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10945070 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17132691 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10945069 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17132695 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17132699 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10945064 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10945060 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10945073 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10945071 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31571873 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10945063 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10945065 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31571883 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31571884 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31571840 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945064 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571885 - -
          Agilent Whole Chr array_18 19 21 A_17_P17132714 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571826 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571840 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571838 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945059 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945051 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571837 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945071 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945057 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945047 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571884 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945066 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571852 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945075 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945050 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945052 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945073 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945053 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945060 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945058 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945046 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571841 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945055 - -
          Agilent Whole Chr array_18 19 21 A_17_P17132699 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571883 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945048 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945074 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945068 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945065 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571876 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945077 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945067 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571864 - -
          Agilent Whole Chr array_18 19 21 A_17_P17132695 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945054 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945072 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571873 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571848 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571844 - -
          Agilent Whole Chr array_18 19 21 A_17_P17132704 - -
          Agilent Whole Chr array_18 19 21 A_17_P17132701 - -
          Agilent Whole Chr array_18 19 21 A_17_P17132688 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571882 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945069 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571842 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945070 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945076 - -
          Agilent Whole Chr array_18 19 21 A_17_P31571846 - -
          Agilent Whole Chr array_18 19 21 A_17_P17132691 - -
          Agilent Whole Chr array_18 19 21 A_17_P17132683 - -
          Agilent Whole Chr array_18 19 21 A_17_P17132687 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945062 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945056 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945049 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945045 - -
          Agilent Whole Chr array_18 19 21 A_17_P10945063 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31571842 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10945063 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10945056 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31571883 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10945069 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31571864 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10945053 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10945067 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10945065 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17132691 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17132695 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10945064 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10945070 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31571852 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31571840 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31571876 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17132699 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P17132701 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31571884 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10945071 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31571846 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10945074 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10945054 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10945073 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31571873 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10945060 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10945059 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10945048 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10945054 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17132701 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10945073 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10945047 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10945051 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10945071 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10945052 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17132695 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10945072 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10945066 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10945045 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10945075 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10945077 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10945064 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10945065 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10945056 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10945058 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10945062 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10945069 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17132690 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17132687 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17132704 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10945060 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10945053 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P10945050 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17132706 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1900010432.hg.1 ENST00000595013.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010432.hg.1 ENST00000595013.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010432.hg.1 ENST00000595013.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010432.hg.1 ENST00000595013.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010432.hg.1 ENST00000597830.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010432.hg.1 ENST00000597830.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010432.hg.1 NR_027620.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900010432.hg.1 NR_027620.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900010432.hg.1 NR_027620.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900010432.hg.1 NR_027620.3 Refseq
          Affymetrix Human Clariom D Assay TC1900010432.hg.1 ENST00000595013.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010432.hg.1 ENST00000595013.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010432.hg.1 ENST00000595013.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010432.hg.1 ENST00000595013.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010432.hg.1 ENST00000597830.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010432.hg.1 ENST00000597830.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010432.hg.1 NR_027620.3 Refseq
          Affymetrix Human Clariom D Assay TC1900010432.hg.1 NR_027620.3 Refseq
          Affymetrix Human Clariom D Assay TC1900010432.hg.1 NR_027620.3 Refseq
          Affymetrix Human Clariom D Assay TC1900010432.hg.1 NR_027620.3 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A011333 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A011333 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1668239 NR_027620.2 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1668239 CD173361.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1668239 NR_027620.2 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1668239 CD173361.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1668239 NR_027620.2 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1668239 CD173361.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1668239 NR_027620.2 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1668239 CD173361.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1668239 NR_027620.2 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1668239 CD173361.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0