Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04233444_s1
          See other PCNAP1 GE Assays ›
          Gene Symbol:
          PCNAP1
          Gene Name:
          proliferating cell nuclear antigen pseudogene 1
          Gene Aliases:
          p1PCNA
          Chromosome Location:
          Chr.4: 99160593 - 99161647 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04233444_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PCNAP1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_028270.1 1 249 103
          GenBank mRNA CR743211.1 - 1 236 103

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC100507053 LOC100507053 Anti-sense Chr. 4 - 99088857 - 99301356 on NCBI Build GRCh38
          Downstream alcohol dehydrogenase 6 (class V) ADH6 ADH-5 41.0kb Sense Chr. 4 - 99202638 - 99219246 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0400011404.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC04001407.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_5248 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_5249 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03297580 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23456397 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23456413 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03297588 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23456414 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23456398 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03297581 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23456402 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23456404 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23456412 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23456410 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23456419 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15595583 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23456416 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23456396 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03297585 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23456417 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03297586 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23456417 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03297580 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23456402 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23456419 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23456414 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23456416 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23456396 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15595583 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03297585 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03297581 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23456404 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03297586 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P03297588 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23456397 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23456413 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23456398 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23456412 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P23456410 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0400011404.hg.1 ENST00000505363.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011404.hg.1 ENST00000505363.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011404.hg.1 ENST00000595299.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011404.hg.1 ENST00000595299.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011404.hg.1 ENST00000595299.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011404.hg.1 NR_028270.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011404.hg.1 NR_028270.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011404.hg.1 NR_028270.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04019625.hg.1 NR_028270.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04019625.hg.1 TC04001407.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0400177969.hg.1 ENST00000505363.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400177969.hg.1 NR_028270.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400177969.hg.1 TC0400011404.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400177969.hg.1 ENST00000505363.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400177969.hg.1 NR_028270.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400177967.hg.1 ENST00000595299.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400177967.hg.1 NR_028270.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400177967.hg.1 TC0400011404.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400177967.hg.1 ENST00000595299.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400177967.hg.1 NR_028270.1 Refseq
          Affymetrix Human Clariom D Assay TC0400011404.hg.1 ENST00000505363.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400011404.hg.1 ENST00000505363.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400011404.hg.1 ENST00000595299.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011404.hg.1 ENST00000595299.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011404.hg.1 ENST00000595299.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011404.hg.1 NR_028270.1 Refseq
          Affymetrix Human Clariom D Assay TC0400011404.hg.1 NR_028270.1 Refseq
          Affymetrix Human Clariom D Assay TC0400011404.hg.1 NR_028270.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400177968.hg.1 ENST00000505363.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400177968.hg.1 ENST00000595299.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400177968.hg.1 NR_028270.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400177968.hg.1 TC0400011404.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400177968.hg.1 ENST00000505363.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400177968.hg.1 ENST00000595299.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400177968.hg.1 NR_028270.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A024518 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A024517 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A024518 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A024517 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-875ns:80/100.66.130.62:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0