Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04233499_s1
          See other SNORA77 GE Assays ›
          Gene Symbol:
          SNORA77
          Gene Name:
          small nucleolar RNA, H/ACA box 77
          Gene Aliases:
          ACA63, SNORA77A
          Chromosome Location:
          Chr.1: 203729581 - 203729705 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04233499_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SNORA77
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003019.1 1 56 93
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DW453250.1 - 1 75 Chr. 1 - 203729576 - 203729749 [+] on Build GRCh38 93

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream opticin OPTC OPT -220.6kb Sense Chr. 1 - 203494143 - 203508949 on NCBI Build GRCh38
          Downstream long intergenic non-protein coding RNA 260 LINC00260 C1orf217,
          NCRNA00260
          872.0 Sense Chr. 1 - 203730577 - 203731853 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100011268.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01001689.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_HS_44k_4047 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800343 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800349 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15164772 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800352 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800325 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00739070 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800328 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00739068 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800367 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800332 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800370 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800329 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800368 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800351 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800354 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00739051 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00739059 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15164774 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00739067 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00739063 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00739077 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15164778 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800326 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00739087 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800335 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00739062 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00739072 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00739056 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15164776 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800365 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800344 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800345 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800324 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00739080 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15164769 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00739050 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00739069 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15164774 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800365 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00739072 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800349 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15164769 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800368 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00739063 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00739051 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00739077 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00739068 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800354 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15164772 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15164776 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800329 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00739050 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00739056 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800351 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15164778 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800335 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00739067 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00739062 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00739070 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00739087 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800367 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00739059 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800345 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800344 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800326 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800325 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800328 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800343 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800352 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00739069 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800324 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800332 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20800370 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00739080 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00739048 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00739055 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00739045 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00739074 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00739052 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15164774 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00739053 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00739061 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00739091 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00739070 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00739050 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00739076 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00739057 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15164772 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00739067 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00739082 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00739087 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15164777 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00739080 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15164781 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00739064 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00739073 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P00739085 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P15164769 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Genome U219 Array 11763897_PM_at - -
          Affymetrix Human Genome U219 Array 11763897_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR01026253.hg.1 ENST00000408716.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01026253.hg.1 NR_003019.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01026253.hg.1 TC01001689.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0100011268.hg.1 ENST00000408716.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011268.hg.1 NR_003019.1 Refseq
          Affymetrix Human Clariom D Assay TC0100011268.hg.1 ENST00000408716.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100011268.hg.1 NR_003019.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100182767.hg.1 ENST00000408716.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100182767.hg.1 NR_003019.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100182767.hg.1 TC0100011268.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100182767.hg.1 NR_003019.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100182767.hg.1 ENST00000408716.1 Ensembl
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3243428 NR_003019.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3243428 NR_003019.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3243428 NR_003019.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3243428 NR_003019.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0