Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04233737_s1
          See other SAP30L-AS1 GE Assays ›
          Gene Symbol:
          SAP30L-AS1
          Gene Name:
          SAP30L antisense RNA 1 (head to head)
          Gene Aliases:
          GALNT10-AS1
          Chromosome Location:
          Chr.5: 154389769 - 154445822 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04233737_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SAP30L-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_037897.1 3 3149 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0500012546.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC05001968.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04385013 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04385008 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24564275 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04385009 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04385011 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24564259 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15754306 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04385018 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P04385010 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24564264 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 157801 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 68744 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 77433 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 304096 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 955530 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 5112 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 11618 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 19567 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 848055 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 732060 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 322562 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 203297 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 125715 AB011263.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 125715 AB020532.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 125715 AB209591.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 125715 AF092032.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 125715 AJ130718.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 125715 AK025377.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 125715 AK309128.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 125715 AK314351.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 125715 AL365450.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 125715 AL365451.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 125715 BC003062.2 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 125715 BC010107.2 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 125715 BX161519.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 125715 BX248291.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 125715 CR601579.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 125715 CR605487.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 125715 CR613648.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 125715 CR617040.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 125715 CR626419.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 125715 NM_003982.2 refseq_mrna_human_NM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 125715 Y18474.2 genbank_mrna_human
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3704280 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 43262 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P55111292 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P37422352 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P17356392 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3704280 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P17356392 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P37422352 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P17356400 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P17356392 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 153916 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 44301 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P17356400 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P17356392 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P37422359 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P17356386 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P37422352 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01370092 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01370070 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01370075 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P37422344 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P17356383 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P17356374 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P17356392 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P17356400 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P37422352 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 220296_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR05027414.hg.1 ENST00000524264.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05027414.hg.1 NR_037897.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05027414.hg.1 TC05001968.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR05027409.hg.1 NR_037897.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05027409.hg.1 TC05001968.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR05027410.hg.1 ENST00000524264.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05027410.hg.1 NR_037897.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05027410.hg.1 TC05001968.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR05027413.hg.1 ENST00000524264.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05027413.hg.1 NR_037897.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05027413.hg.1 TC05001968.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0500187603.hg.1 ENST00000524264.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500187603.hg.1 ENST00000524264.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500187603.hg.1 NR_037897.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500187603.hg.1 TC0500012546.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500187603.hg.1 NR_037897.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500187603.hg.1 ENST00000524264.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500187603.hg.1 ENST00000524264.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500187601.hg.1 NR_037897.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500187601.hg.1 TC0500012546.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500187601.hg.1 NR_037897.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500187604.hg.1 ENST00000524264.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500187604.hg.1 ENST00000524264.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500187604.hg.1 NR_037897.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500187604.hg.1 TC0500012546.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500187604.hg.1 ENST00000524264.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500187604.hg.1 ENST00000524264.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500187604.hg.1 NR_037897.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500187611.hg.1 ENST00000501280.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0500187611.hg.1 ENST00000519727.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500187611.hg.1 ENST00000519727.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500187611.hg.1 ENST00000522312.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500187611.hg.1 ENST00000524264.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500187611.hg.1 ENST00000524264.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500187611.hg.1 NR_037897.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500187611.hg.1 TC0500012546.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500187611.hg.1 ENST00000501280.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0500187611.hg.1 ENST00000519727.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500187611.hg.1 ENST00000519727.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500187611.hg.1 ENST00000522312.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500187611.hg.1 ENST00000524264.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500187611.hg.1 ENST00000524264.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500187611.hg.1 NR_037897.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500187602.hg.1 NR_037897.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500187602.hg.1 TC0500012546.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500187602.hg.1 NR_037897.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500187612.hg.1 ENST00000501280.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0500187612.hg.1 ENST00000519727.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500187612.hg.1 ENST00000519727.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500187612.hg.1 ENST00000522312.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500187612.hg.1 ENST00000524264.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500187612.hg.1 ENST00000524264.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500187612.hg.1 NR_037897.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500187612.hg.1 TC0500012546.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500187612.hg.1 ENST00000501280.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0500187612.hg.1 ENST00000519727.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500187612.hg.1 ENST00000519727.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500187612.hg.1 ENST00000522312.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500187612.hg.1 ENST00000524264.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500187612.hg.1 ENST00000524264.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500187612.hg.1 NR_037897.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A027614 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A027614 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 204400 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0