Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04234270_g1
          See other KCNH2 GE Assays ›
          Gene Symbol:
          KCNH2
          Gene Name:
          potassium voltage-gated channel subfamily H member 2
          Gene Aliases:
          ERG-1, ERG1, H-ERG, HERG, HERG1, Kv11.1, LQT2, SQT1
          Chromosome Location:
          Chr.7: 150944956 - 150978314 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04234270_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: KCNH2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000238.3 NP_000229.1 6 - 7 1955 98
          NM_001204798.1 NP_001191727.1 2 - 3 860 98
          NM_172056.2 NP_742053.1 6 - 7 1955 98
          NM_172057.2 NP_742054.1 2 - 3 860 98
          XM_011516185.2 XP_011514487.1 5 - 6 1366 98
          XM_011516186.2 XP_011514488.1 6 - 7 1947 98
          XM_017012195.1 XP_016867684.1 5 - 6 2036 98
          XM_017012196.1 XP_016867685.1 6 - 7 1496 98
          GenBank mRNA AB044806.1 - 6 - 7 1737 98
          AF363636.1 - 6 - 7 1554 98
          AJ512214.1 - 2 - 3 551 98
          AJ609614.1 - 2 - 3 534 98
          BC001914.1 - 5 - 6 1211 98
          BC127673.1 - 2 - 3 849 98
          BT007336.1 - 5 - 6 1206 98
          DQ525913.1 - 6 - 7 1955 98
          FJ938021.1 - 4 - 5 2387 98
          U04270.1 - 6 - 7 1737 98
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BC008341.1 - 908 Chr. - - [-] on Build 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700013050.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07002024.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene KCNH2

          Panther Classification:

          Molecular Function -

          ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          phosphorelay signal transduction system
          regulation of heart rate by hormone
          signal transduction by protein phosphorylation
          cellular response to drug
          regulation of membrane potential
          potassium ion homeostasis
          cardiac muscle contraction
          regulation of membrane repolarization
          regulation of ventricular cardiac muscle cell membrane repolarization
          cardiac conduction
          potassium ion export
          potassium ion transmembrane transport
          ventricular cardiac muscle cell action potential
          membrane depolarization during action potential
          membrane repolarization during action potential
          membrane repolarization during cardiac muscle cell action potential
          regulation of heart rate by cardiac conduction
          potassium ion export across plasma membrane
          membrane repolarization during ventricular cardiac muscle cell action potential
          regulation of potassium ion transmembrane transport
          negative regulation of potassium ion transmembrane transport
          positive regulation of potassium ion transmembrane transport
          negative regulation of potassium ion export
          phosphorelay sensor kinase activity
          inward rectifier potassium channel activity
          voltage-gated potassium channel activity
          delayed rectifier potassium channel activity
          protein binding
          ubiquitin protein ligase binding
          identical protein binding
          protein homodimerization activity
          C3HC4-type RING finger domain binding
          voltage-gated potassium channel activity involved in cardiac muscle cell action potential repolarization
          scaffold protein binding
          voltage-gated potassium channel activity involved in ventricular cardiac muscle cell action potential repolarization

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000262186.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000262186.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000262186.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000262186.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000262186.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000262186.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000262186.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000262186.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000262186.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000262186.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000262186.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000262186.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000262186.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000262186.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000262186.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000262186.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000262186.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000262186.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000262186.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000262186.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000262186.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000262186.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000262186.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000330883.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000330883.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000330883.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000330883.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000330883.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000330883.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000330883.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000330883.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000330883.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000330883.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000330883.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000330883.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000330883.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000330883.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000430723.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000430723.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000430723.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000430723.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000430723.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000430723.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000430723.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000430723.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000430723.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000430723.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000430723.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000430723.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000430723.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000461280.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000461280.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000461280.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000461280.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000461280.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000461280.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000461280.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000461280.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000473610.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000473610.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000473610.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000473610.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000473610.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000473610.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000473610.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000473610.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000473610.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000532957.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000532957.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000532957.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000532957.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000532957.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000532957.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000532957.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000532957.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000532957.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000532957.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000532957.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000532957.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000532957.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000532957.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000532957.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000532957.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 ENST00000532957.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_000238.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_000238.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_000238.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_000238.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_000238.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_000238.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_000238.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_000238.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_000238.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_000238.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_000238.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_000238.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_000238.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_000238.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_000238.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_000238.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_000238.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_000238.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_000238.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_000238.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_000238.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_000238.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_000238.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_001204798.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_001204798.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_001204798.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_001204798.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_001204798.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_001204798.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_001204798.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_001204798.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_172056.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_172056.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_172056.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_172056.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_172056.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_172056.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_172056.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_172056.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_172056.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_172056.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_172056.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_172056.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_172056.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_172056.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_172056.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_172056.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_172056.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_172056.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_172057.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_172057.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_172057.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_172057.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_172057.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_172057.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_172057.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_172057.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_172057.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_172057.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_172057.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_172057.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013050.hg.1 NM_172057.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 223860 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0