Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04259676_g1
          See other FSCN1 GE Assays ›
          Gene Symbol:
          FSCN1
          Gene Name:
          fascin actin-bundling protein 1
          Gene Aliases:
          FAN1, HSN, SNL, p55
          Chromosome Location:
          Chr.7: 5592805 - 5606656 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04259676_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: FSCN1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_003088.3 NP_003079.1 1 - 2 960 63
          GenBank mRNA AK095254.1 - 2 - 3 894 63
          AK316607.1 - 1 - 2 955 63
          BC000521.2 - 1 - 2 851 2961047 63
          BC006304.2 - 1 - 2 942 4100996 63
          BC007539.1 - 1 - 2 851 63
          BC007643.1 - 1 - 2 584 63
          BC007948.2 - 1 - 2 939 4299524 63
          BC007988.2 - 1 - 2 851 2961047 63
          BT006636.1 - 1 - 2 828 63
          U03057.1 - 1 - 2 939 63
          U09873.1 - 1 - 2 873 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700006629.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07000055.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene FSCN1

          Gene Ontology Categories:

          Function(s) Process(es)

          cell-cell junction assembly
          cell population proliferation
          anatomical structure morphogenesis
          positive regulation of lamellipodium assembly
          cell migration
          microspike assembly
          actin cytoskeleton organization
          regulation of microvillus assembly
          regulation of actin cytoskeleton organization
          establishment of apical/basal cell polarity
          cell motility
          actin filament bundle assembly
          positive regulation of filopodium assembly
          positive regulation of podosome assembly
          positive regulation of extracellular matrix disassembly
          cell-cell adhesion
          RNA binding
          actin binding
          protein binding
          protein-macromolecule adaptor activity
          actin filament binding
          cadherin binding involved in cell-cell adhesion

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR07000888.hg.1 ENST00000340250.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07000888.hg.1 ENST00000382361.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07000888.hg.1 NM_003088.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07000888.hg.1 TC07000055.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000382361.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000382361.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000382361.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000382361.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000382361.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000382361.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000382361.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000382361.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000382361.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000382361.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000382361.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000382361.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000405801.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000405801.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000405801.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000405801.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000405801.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000444748.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000444748.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000444748.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000444748.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000444748.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000447103.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000447103.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000447103.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000447103.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000447103.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000447103.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000473330.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000473330.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000473330.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000473330.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000473330.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000473330.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000473330.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000473330.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000473330.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000473330.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 ENST00000473330.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 NM_003088.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 NM_003088.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 NM_003088.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 NM_003088.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 NM_003088.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 NM_003088.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 NM_003088.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 NM_003088.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 NM_003088.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 NM_003088.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 NM_003088.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700006629.hg.1 NM_003088.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07000887.hg.1 ENST00000382361.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07000887.hg.1 NM_003088.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07000887.hg.1 TC07000055.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07000885.hg.1 ENST00000340250.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07000885.hg.1 ENST00000382361.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07000885.hg.1 NM_003088.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07000885.hg.1 TC07000055.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07000886.hg.1 ENST00000340250.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07000886.hg.1 ENST00000382361.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07000886.hg.1 NM_003088.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07000886.hg.1 TC07000055.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1500006827.hg.1 ENST00000568912.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0700145583.hg.1 ENST00000382361.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0700145583.hg.1 NM_003088.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700145583.hg.1 TC0700006629.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700145583.hg.1 NM_003088.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700145583.hg.1 ENST00000382361.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0700145585.hg.1 ENST00000382361.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0700145585.hg.1 NM_003088.3 Refseq
          Affymetrix Human Clariom D Assay PSR0700145585.hg.1 TC0700006629.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700145585.hg.1 ENST00000382361.7 Ensembl
          Affymetrix Human Clariom D Assay PSR0700145585.hg.1 NM_003088.3 Refseq
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000382361.7 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000382361.7 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000382361.7 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000382361.7 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000382361.7 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000382361.7 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000382361.7 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000382361.7 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000382361.7 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000382361.7 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000382361.7 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000382361.7 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000405801.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000405801.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000405801.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000405801.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000405801.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000444748.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000444748.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000444748.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000444748.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000444748.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000447103.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000447103.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000447103.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000447103.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000447103.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000447103.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000473330.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000473330.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000473330.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000473330.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000473330.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000473330.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000473330.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000473330.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000473330.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000473330.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 ENST00000473330.1 Ensembl
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 NM_003088.3 Refseq
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 NM_003088.3 Refseq
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 NM_003088.3 Refseq
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 NM_003088.3 Refseq
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 NM_003088.3 Refseq
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 NM_003088.3 Refseq
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 NM_003088.3 Refseq
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 NM_003088.3 Refseq
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 NM_003088.3 Refseq
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 NM_003088.3 Refseq
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 NM_003088.3 Refseq
          Affymetrix Human Clariom D Assay TC0700006629.hg.1 NM_003088.3 Refseq
          Affymetrix Human Clariom D Assay TC1500006827.hg.1 ENST00000568912.1 Ensembl
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 150201 - -
          ABI Human Genome Survey Microarray Version 2.0 169875 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0