Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04273344_s1
          See other MIR641 GE Assays ›
          Gene Symbol:
          MIR641
          Gene Name:
          microRNA 641
          Gene Aliases:
          MIRN641, hsa-mir-641, mir-641
          Chromosome Location:
          Chr.19: 40282543 - 40282641 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04273344_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MIR641
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_030371.1 1 45 96

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream cyclin N-terminal domain containing 2 CNTD2 CCNP -55.4kb Sense Chr. 19 - 40222208 - 40227111 on NCBI Build GRCh38
          Downstream chromosome 19 open reading frame 47 C19orf47 12.6kb Sense Chr. 19 - 40292743 - 40348527 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
          This assay targets the same location as Clariom D transcript cluster(s) TC1900010697.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC19001534.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Chr array_18 19 21 A_17_P31591579 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961586 - -
          Agilent Whole Chr array_18 19 21 A_17_P31591582 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961582 - -
          Agilent Whole Chr array_18 19 21 A_17_P31591592 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961583 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961571 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961585 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961578 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961587 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961575 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961572 - -
          Agilent Whole Chr array_18 19 21 A_17_P31591584 - -
          Agilent Whole Chr array_18 19 21 A_17_P31591587 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961570 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961581 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961588 - -
          Agilent Whole Chr array_18 19 21 A_17_P31591580 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961576 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961573 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961574 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961584 - -
          Agilent Whole Chr array_18 19 21 A_17_P31591581 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961579 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961577 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961580 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10961571 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10961583 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10961580 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10961577 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31591592 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31591580 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10961575 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10961573 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10961570 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10961584 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961588 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961586 - -
          Agilent Whole Chr array_18 19 21 A_17_P31591582 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961578 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961583 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961580 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961579 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961576 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961585 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961582 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961587 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961574 - -
          Agilent Whole Chr array_18 19 21 A_17_P31591584 - -
          Agilent Whole Chr array_18 19 21 A_17_P31591581 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961575 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961581 - -
          Agilent Whole Chr array_18 19 21 A_17_P31591579 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961571 - -
          Agilent Whole Chr array_18 19 21 A_17_P31591587 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961572 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961584 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961577 - -
          Agilent Whole Chr array_18 19 21 A_17_P31591580 - -
          Agilent Whole Chr array_18 19 21 A_17_P31591592 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961573 - -
          Agilent Whole Chr array_18 19 21 A_17_P10961570 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10961570 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10961583 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10961584 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10961575 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31591580 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10961573 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10961571 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10961577 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P10961580 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31591592 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR19020955.hg.1 TC19001534.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19020955.hg.1 ENST00000384899.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR19020955.hg.1 NR_030371.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900010697.hg.1 ENST00000384899.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010697.hg.1 NR_030371.1 Refseq
          Affymetrix Human Clariom D Assay TC1900010697.hg.1 ENST00000384899.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010697.hg.1 NR_030371.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900188406.hg.1 ENST00000384899.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900188406.hg.1 NR_030371.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900188406.hg.1 TC1900010697.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900188406.hg.1 NR_030371.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900188406.hg.1 ENST00000384899.1 Ensembl
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3308190 NR_030371.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3308190 NR_030371.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3308190 NR_030371.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3308190 NR_030371.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0