Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04273357_s1
          See other MIR549A GE Assays ›
          Gene Symbol:
          MIR549A
          Gene Name:
          microRNA 549a
          Gene Aliases:
          MIR549, MIRN549, hsa-mir-549, hsa-mir-549a, mir-549a
          Chromosome Location:
          Chr.15: 80841978 - 80842073 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04273357_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MIR549A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_030393.1 1 49 89

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream abhydrolase domain containing 17C ABHD17C FAM108C1 -86.4kb Anti-sense Chr. 15 - 80695311 - 80755621 on NCBI Build GRCh38
          Downstream mesoderm development candidate 2 MESDC2 BOCA,
          MESD
          105.3kb Sense Chr. 15 - 80947326 - 80989878 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
          This assay targets the same location as Clariom D transcript cluster(s) TC1500010200.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC15001726.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785949 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785955 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785956 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785953 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283558 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785930 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785943 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785909 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283518 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283531 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283519 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785935 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283548 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785920 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283529 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283566 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283536 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785948 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283520 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283570 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283537 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785925 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785931 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785916 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283553 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283535 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785946 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283550 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283549 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785961 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785927 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785952 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785951 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785911 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785921 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785915 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283574 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785941 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785938 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785933 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283571 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785913 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283571 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785938 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283537 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785920 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283549 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785915 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283519 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785921 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283529 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785913 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785933 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283553 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785909 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283518 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283574 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283548 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785955 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785935 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785925 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785927 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785952 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785949 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785916 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785946 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785951 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785941 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785956 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283520 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785961 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283535 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785948 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283570 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283550 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785953 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283566 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785931 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283558 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785930 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283531 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785911 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P30283536 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P09785943 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR15016885.hg.1 ENST00000385268.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15016885.hg.1 NR_030393.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15016885.hg.1 TC15001726.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1500010200.hg.1 ENST00000385268.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500010200.hg.1 NR_030393.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500176571.hg.1 ENST00000385268.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500176571.hg.1 NR_030393.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500176571.hg.1 TC1500010200.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500176571.hg.1 NR_030393.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500176571.hg.1 ENST00000385268.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500010200.hg.1 ENST00000385268.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500010200.hg.1 NR_030393.1 Refseq
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3309899 NR_030393.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3309899 NR_030393.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3309899 NR_030393.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3309899 NR_030393.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0