Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04273366_s1
          See other MIR765 GE Assays ›
          Gene Symbol:
          MIR765
          Gene Name:
          microRNA 765
          Gene Aliases:
          MIRN765, hsa-mir-765, mir-765
          Chromosome Location:
          Chr.1: 156936131 - 156936244 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04273366_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MIR765
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_030527.1 1 52 77
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AA054058.1 - 2 398 Chr. 1 - 156936135 - 156991697 [-] on Build GRCh38 77

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream leucine rich repeat containing 71 LRRC71 C1orf92 Anti-sense Chr. 1 - 156920608 - 156941208 on NCBI Build GRCh38
          Downstream ETS variant 3 like ETV3L 155.8kb Sense Chr. 1 - 157092043 - 157099808 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
          This assay targets the same location as Clariom D transcript cluster(s) TC0100016028.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01003357.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530806 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530864 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530848 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20589217 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530855 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530841 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530840 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530799 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530842 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530814 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530828 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530850 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530844 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530843 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530867 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530833 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20589242 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530865 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530857 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15133727 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20589218 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20589228 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20589254 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530859 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530809 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20589256 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530846 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20589250 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530830 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20589192 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530817 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530851 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530854 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20589197 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530819 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530860 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530831 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20589219 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20589232 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530815 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530853 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530807 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530800 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20589253 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530798 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530836 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530803 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530821 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530813 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20589216 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20589219 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530848 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530850 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20589228 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530809 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530831 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530855 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530799 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20589250 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530854 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530860 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20589218 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530807 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530840 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530867 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20589253 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530819 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530806 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20589232 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20589242 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530843 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530815 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530833 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20589254 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530813 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530859 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20589256 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530814 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530830 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530864 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530846 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530857 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530798 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530844 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530851 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20589216 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530817 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530800 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530828 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20589217 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530841 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530821 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530865 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530836 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15133727 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530842 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530853 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00530803 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20589192 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20589197 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100016028.hg.1 ENST00000390226.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100016028.hg.1 NR_030527.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01052507.hg.1 ENST00000390226.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01052507.hg.1 NR_030527.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01052507.hg.1 TC01003357.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0100016028.hg.1 ENST00000390226.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100016028.hg.1 NR_030527.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100221740.hg.1 ENST00000390226.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100221740.hg.1 NR_030527.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100221740.hg.1 TC0100016028.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100221740.hg.1 NR_030527.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100221740.hg.1 ENST00000390226.1 Ensembl
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3308570 NR_030527.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3308570 NR_030527.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3308570 NR_030527.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3308570 NR_030527.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0