Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04273772_m1
          See other DKFZP451B082 GE Assays ›
          Gene Symbol:
          DKFZp451B082
          Gene Name:
          uncharacterized LOC401282
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 163338342 - 163347033 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04273772_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DKFZp451B082
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033862.1 3 - 4 3740 87
          GenBank mRNA AL831931.1 - 3 - 4 3739 87

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream PACRG antisense RNA 1 PACRG-AS1 -13.9kb Sense Chr. 6 - 163309985 - 163324473 on NCBI Build GRCh38
          Downstream colon adenocarcinoma hypermethylated (non-protein coding) CAHM LINC00468 69.5kb Sense Chr. 6 - 163413065 - 163413950 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0600013816.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC06002291.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226673 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226765 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226710 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226687 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226694 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431962 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226700 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431856 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226776 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226788 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431837 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431961 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226741 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431864 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226668 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431958 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226791 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431869 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226777 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226690 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431897 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226688 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226778 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226724 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431832 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431917 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226755 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226745 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15886376 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226805 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226783 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226698 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431932 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431843 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226672 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226671 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226664 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226740 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226807 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226680 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226683 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226739 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431846 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431853 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226754 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431973 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431963 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431946 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226742 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431960 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226692 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226726 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226751 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226760 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226767 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431857 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226812 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226695 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226744 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431873 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226725 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226781 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226732 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226801 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226663 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431937 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226730 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431884 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226737 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226665 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226786 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226704 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226721 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431841 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226785 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431904 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226686 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226753 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226746 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226799 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226716 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431941 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15886374 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226709 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226771 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431844 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226766 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431974 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431956 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226752 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431928 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431878 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226789 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226808 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226699 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226775 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431868 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226723 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431883 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226750 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226806 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226764 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226736 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431912 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226802 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226684 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226748 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226719 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226712 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05226749 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P25431865 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 252158 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 819552 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 134605 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 775239 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3826751 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 4490 - -
          Agilent Human Genome CGH + SNP Microarray A_18_P15936660 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3826751 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3826751 - -
          Custom Human Agilent Array 11k, Chromosome 6, Expression 906 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01632388 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P15936660 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P17803198 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01632391 - -
          Agilent Human 1B Microarray G4111A A_32_P852920 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P01632388 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P15936660 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1563511_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR06028770.hg.1 NR_033862.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR06028770.hg.1 TC06002291.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0600195343.hg.1 NR_033862.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600195343.hg.1 TC0600013816.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600195343.hg.1 NR_033862.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600195345.hg.1 NR_033862.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600195345.hg.1 TC0600013816.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600195345.hg.1 NR_033862.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600195342.hg.1 NR_033862.1 Refseq
          Affymetrix Human Clariom D Assay PSR0600195342.hg.1 TC0600013816.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0600195342.hg.1 NR_033862.1 Refseq
          Human Genome U133 Plus 2.0 Array 1563511_at - -
          Human Genome U133 Plus 2.0 Array 1563511_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563511_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563511_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563511_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563511_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563511_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563511_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563511_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563511_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563511_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563511_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1563511_at9 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A030795 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A030795 - -
          Array Name Probe IDs Targets Target Type
          Illumina human-6 v2.0 expression beadchip ILMN_1891287 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip ILMN_92861 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_92861 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1891287 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1891287 - -
          Illumina HumanWG-6_V2_0_R2 ILMN_1891287 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-47c8f:80/100.66.130.62:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0