Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04273798_m1
          See other MLK7-AS1 GE Assays ›
          Gene Symbol:
          MLK7-AS1
          Gene Name:
          MLK7 antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 173197712 - 173282036 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04273798_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MLK7-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033882.1 7 - 8 1424 83
          GenBank mRNA BC040565.1 - 7 - 8 1434 83

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream sterile alpha motif and leucine zipper containing kinase AZK ZAK AZK,
          MLK7,
          MLT,
          MLTK,
          MRK,
          SFMMP,
          mlklak,
          pk
          Anti-sense Chr. 2 - 173075435 - 173268009 on NCBI Build GRCh38
          Downstream cell division cycle associated 7 CDCA7 ICF3,
          JPO1
          146.4kb Anti-sense Chr. 2 - 173354017 - 173368992 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200014938.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02002532.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21805511 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21805521 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01681479 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01681489 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21805542 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21805515 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21805519 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01681505 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01681508 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01681471 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01681478 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01681476 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01681501 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21805551 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21805524 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01681519 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21805548 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21805538 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01681496 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21805532 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01681512 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21805545 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01681480 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21805534 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21805506 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21805493 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01681522 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21805517 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21805495 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21805512 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21805535 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01681504 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01681483 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21805529 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21805543 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21805541 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21805499 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21805550 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01681485 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01681493 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 91761 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 176939 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 73610 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 38264 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 653144 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3696837 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 27364 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3696837 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3696837 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P15931870 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P35963631 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15931870 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15931878 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00530882 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15931861 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15931874 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P30771785 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P15931870 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P35963631 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P30771785 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1558653_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR02040156.hg.1 ENST00000423106.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02040156.hg.1 NR_033882.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02040156.hg.1 TC02002532.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02040158.hg.1 ENST00000422703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02040158.hg.1 ENST00000423106.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02040158.hg.1 NR_033882.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02040158.hg.1 TC02002532.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 ENST00000419609.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 ENST00000419609.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 ENST00000419609.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 ENST00000419609.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 ENST00000419609.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 ENST00000419609.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 ENST00000419609.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 ENST00000419609.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 ENST00000422703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 ENST00000422703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 ENST00000422703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 ENST00000422703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 ENST00000422703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 ENST00000422703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 ENST00000422703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 ENST00000422703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 ENST00000422703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 ENST00000422703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 ENST00000423106.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 ENST00000423106.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 ENST00000423106.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 ENST00000423106.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 ENST00000423106.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 ENST00000423106.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 ENST00000423106.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 ENST00000423106.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 ENST00000423106.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 NR_033882.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 NR_033882.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 NR_033882.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 NR_033882.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 NR_033882.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 NR_033882.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 NR_033882.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 NR_033882.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200014938.hg.1 NR_033882.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02040157.hg.1 ENST00000419609.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02040157.hg.1 ENST00000422703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02040157.hg.1 ENST00000423106.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02040157.hg.1 NR_033882.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02040157.hg.1 TC02002532.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0200209162.hg.1 ENST00000422703.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200209162.hg.1 ENST00000422703.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200209162.hg.1 ENST00000423106.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200209162.hg.1 NR_033882.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200209162.hg.1 TC0200014938.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200209162.hg.1 NR_033882.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200209162.hg.1 ENST00000422703.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200209162.hg.1 ENST00000422703.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200209162.hg.1 ENST00000423106.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200209161.hg.1 ENST00000419609.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200209161.hg.1 ENST00000419609.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200209161.hg.1 ENST00000422703.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200209161.hg.1 ENST00000422703.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200209161.hg.1 ENST00000423106.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200209161.hg.1 NR_033882.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200209161.hg.1 TC0200014938.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200209161.hg.1 ENST00000419609.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200209161.hg.1 ENST00000419609.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200209161.hg.1 ENST00000422703.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200209161.hg.1 ENST00000422703.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200209161.hg.1 ENST00000423106.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200209161.hg.1 NR_033882.1 Refseq
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 ENST00000419609.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 ENST00000419609.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 ENST00000419609.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 ENST00000419609.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 ENST00000419609.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 ENST00000419609.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 ENST00000419609.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 ENST00000419609.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 ENST00000422703.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 ENST00000422703.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 ENST00000422703.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 ENST00000422703.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 ENST00000422703.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 ENST00000422703.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 ENST00000422703.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 ENST00000422703.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 ENST00000422703.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 ENST00000422703.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 ENST00000423106.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 ENST00000423106.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 ENST00000423106.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 ENST00000423106.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 ENST00000423106.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 ENST00000423106.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 ENST00000423106.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 ENST00000423106.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 ENST00000423106.2 Ensembl
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 NR_033882.1 Refseq
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 NR_033882.1 Refseq
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 NR_033882.1 Refseq
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 NR_033882.1 Refseq
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 NR_033882.1 Refseq
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 NR_033882.1 Refseq
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 NR_033882.1 Refseq
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 NR_033882.1 Refseq
          Affymetrix Human Clariom D Assay TC0200014938.hg.1 NR_033882.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200209160.hg.1 ENST00000423106.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200209160.hg.1 NR_033882.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200209160.hg.1 TC0200014938.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200209160.hg.1 ENST00000423106.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200209160.hg.1 NR_033882.1 Refseq
          Human Genome U133 Plus 2.0 Array 1558653_PM_at - -
          Human Genome U133 Plus 2.0 Array 1558653_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558653_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558653_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558653_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558653_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558653_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558653_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558653_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558653_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558653_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558653_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1558653_at4 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A014632 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A014632 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-786bfc498d-tbs5g:80/100.66.134.240:80.
          git-commit: 9eb01563f6748445c58f67612c67ecf6b1da7421
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0