Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04273820_m1
          See other MED15P9 GE Assays ›
          Gene Symbol:
          MED15P9
          Gene Name:
          mediator complex subunit 15 pseudogene 9
          Gene Aliases:
          CCDC74B-AS1, MED15P2
          Chromosome Location:
          Chr.2: 130129623 - 130139413 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04273820_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MED15P9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033903.1 1 - 2 173 105

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream POTE ankyrin domain family member F POTEF A26C1B,
          ACTB,
          POTE2alpha,
          POTEACTIN
          -401.0 Anti-sense Chr. 2 - 130073535 - 130129222 on NCBI Build GRCh38
          Downstream coiled-coil domain containing 74B CCDC74B 3.7kb Anti-sense Chr. 2 - 130139283 - 130145134 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200009279.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02000802.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 499512 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 592164 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 562651 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 487913 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 155460 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 108303 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 244798 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1561433_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR02012383.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02012383.hg.1 NR_033903.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02012383.hg.1 TC02000802.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0200009279.hg.1 ENST00000424716.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009279.hg.1 ENST00000424716.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009279.hg.1 ENST00000424716.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009279.hg.1 ENST00000424716.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009279.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009279.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009279.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009279.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009279.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009279.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009279.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0200009279.hg.1 NR_033903.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009279.hg.1 NR_033903.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009279.hg.1 NR_033903.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009279.hg.1 NR_033903.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009279.hg.1 NR_033903.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009279.hg.1 NR_033903.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0200009279.hg.1 NR_033903.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02012385.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02012385.hg.1 NR_033903.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02012385.hg.1 TC02000802.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02012384.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02012384.hg.1 NR_033903.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02012384.hg.1 TC02000802.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02012387.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02012387.hg.1 NR_033903.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02012387.hg.1 TC02000802.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02012388.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02012388.hg.1 NR_033903.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02012388.hg.1 TC02000802.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02012386.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02012386.hg.1 NR_033903.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02012386.hg.1 TC02000802.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0200164950.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200164950.hg.1 NR_033903.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200164950.hg.1 TC0200009279.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200164950.hg.1 NR_033903.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200164950.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200164948.hg.1 ENST00000424716.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200164948.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200164948.hg.1 NR_033903.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200164948.hg.1 TC0200009279.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200164948.hg.1 ENST00000424716.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200164948.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200164948.hg.1 NR_033903.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200164942.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200164942.hg.1 NR_033903.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200164942.hg.1 TC0200009279.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200164942.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200164942.hg.1 NR_033903.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200164943.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200164943.hg.1 NR_033903.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200164943.hg.1 TC0200009279.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200164943.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200164943.hg.1 NR_033903.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200164949.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200164949.hg.1 NR_033903.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200164949.hg.1 TC0200009279.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200164949.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200164949.hg.1 NR_033903.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200164951.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200164951.hg.1 NR_033903.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200164951.hg.1 TC0200009279.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200164951.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200164951.hg.1 NR_033903.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009279.hg.1 ENST00000424716.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009279.hg.1 ENST00000424716.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009279.hg.1 ENST00000424716.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009279.hg.1 ENST00000424716.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009279.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009279.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009279.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009279.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009279.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009279.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009279.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Clariom D Assay TC0200009279.hg.1 NR_033903.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009279.hg.1 NR_033903.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009279.hg.1 NR_033903.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009279.hg.1 NR_033903.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009279.hg.1 NR_033903.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009279.hg.1 NR_033903.1 Refseq
          Affymetrix Human Clariom D Assay TC0200009279.hg.1 NR_033903.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200164946.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200164946.hg.1 NR_033903.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200164946.hg.1 TC0200009279.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200164946.hg.1 ENST00000427638.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200164946.hg.1 NR_033903.1 Refseq
          Human Genome U133 Plus 2.0 Array 1561433_PM_at - -
          Human Genome U133 Plus 2.0 Array 1561433_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561433_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561433_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561433_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561433_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561433_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561433_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561433_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561433_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561433_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561433_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561433_at1 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A016334 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A016334 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0