Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04273851_g1
          See other LINC00884 GE Assays ›
          Gene Symbol:
          LINC00884
          Gene Name:
          long intergenic non-protein coding RNA 884
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 194487140 - 194488545 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04273851_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC00884
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033929.1 1 - 2 31 96

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ATPase 13A3 ATP13A3 AFURS1 -486.0 Anti-sense Chr. 3 - 194402674 - 194487006 on NCBI Build GRCh38
          Downstream uncharacterized LOC105374290 LOC105374290 10.5kb Anti-sense Chr. 3 - 194499007 - 194527436 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0300009933.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC03001074.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02872420 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02872438 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012819 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012822 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012934 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15524492 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012797 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02872543 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012824 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15524516 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012787 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02872549 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012863 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02872455 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02872422 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02872462 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012781 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012820 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02872559 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02872454 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02872530 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012942 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012825 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02872442 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012803 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02872527 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012804 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012859 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012953 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012937 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012848 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012847 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02872461 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012941 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15524511 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02872469 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012801 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012849 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15524493 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012783 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012806 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012957 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012798 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012794 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02872475 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02872410 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02872561 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012839 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012955 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02872411 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012828 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02872531 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02872409 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02872533 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012810 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012928 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012916 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012779 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012939 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02872551 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02872468 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012861 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012949 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012816 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012792 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012782 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02872532 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012778 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02872443 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012807 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012923 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02872476 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012862 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02872401 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012776 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02872535 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012800 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02872437 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012948 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012788 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012944 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02872529 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012914 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02872540 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012823 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012867 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012791 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02872550 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02872536 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012818 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23012815 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 31215 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 122881 AF057705.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 122881 BC005956.1 genbank_mrna_human
          Agilent SurePrint G3 Human CGH Microarray 4x180K 122881 NM_006911.2 refseq_mrna_human_NM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 73322 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 574200 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 10302 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 592337 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 186678 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 649451 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 943454 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 706782 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 219950 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 366009 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3390177 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 52496 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3390177 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3390177 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P16555448 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P16555433 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P16555448 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P00901588 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 14633 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 26 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 81027 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P16555448 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00901620 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P14563687 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00901588 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P14566787 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00901607 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P36604427 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00901605 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P31197057 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00901597 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P16555433 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P16555456 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00901586 - -
          Agilent Human 1B Microarray G4111A A_32_P335910 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P36604427 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P00901588 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P14566787 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P31197057 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P16555456 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P00901620 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P16555448 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1566761_PM_a_at - -
          Affymetrix HT HG-U133+ PM Array Plate 1566760_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR03018722.hg.1 NR_033929.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03018722.hg.1 TC03001074.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03018724.hg.1 ENST00000448892.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03018724.hg.1 NR_033929.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03018724.hg.1 TC03001074.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0300009933.hg.1 ENST00000437597.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009933.hg.1 ENST00000437597.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009933.hg.1 ENST00000437597.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009933.hg.1 ENST00000448892.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009933.hg.1 ENST00000448892.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009933.hg.1 ENST00000448892.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009933.hg.1 ENST00000448892.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009933.hg.1 ENST00000448892.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009933.hg.1 ENST00000448892.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009933.hg.1 ENST00000448892.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300009933.hg.1 NR_033929.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009933.hg.1 NR_033929.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300009933.hg.1 NR_033929.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300172158.hg.1 ENST00000448892.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300172158.hg.1 NR_033929.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300172158.hg.1 TC0300009933.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300172158.hg.1 NR_033929.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300172158.hg.1 ENST00000448892.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009933.hg.1 ENST00000437597.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009933.hg.1 ENST00000437597.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009933.hg.1 ENST00000437597.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009933.hg.1 ENST00000448892.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009933.hg.1 ENST00000448892.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009933.hg.1 ENST00000448892.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009933.hg.1 ENST00000448892.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009933.hg.1 ENST00000448892.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009933.hg.1 ENST00000448892.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009933.hg.1 ENST00000448892.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300009933.hg.1 NR_033929.1 Refseq
          Affymetrix Human Clariom D Assay TC0300009933.hg.1 NR_033929.1 Refseq
          Affymetrix Human Clariom D Assay TC0300009933.hg.1 NR_033929.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300172159.hg.1 ENST00000448892.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300172159.hg.1 NR_033929.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300172159.hg.1 TC0300009933.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300172159.hg.1 ENST00000448892.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300172159.hg.1 NR_033929.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300172155.hg.1 NR_033929.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300172155.hg.1 TC0300009933.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300172155.hg.1 NR_033929.1 Refseq
          Human Genome U133 Plus 2.0 Array 1566760_at - -
          Human Genome U133 Plus 2.0 Array 242853_PM_at - -
          Human Genome U133 Plus 2.0 Array 1566761_PM_a_at - -
          Human Genome U133 Plus 2.0 Array 1566761_a_at - -
          Human Genome U133 Plus 2.0 Array 242853_at - -
          Human Genome U133 Plus 2.0 Array 1566760_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566761_a_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566760_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566761_a_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566761_a_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566760_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566761_a_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566760_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566760_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566761_a_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566761_a_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566761_a_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566761_a_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566760_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566760_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566761_a_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566760_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566760_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566761_a_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566760_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566761_a_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566760_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1566760_at10 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A023736 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A023736 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0