Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04273871_m1
          See other SCOC-AS1 GE Assays ›
          Gene Symbol:
          SCOC-AS1
          Gene Name:
          SCOC antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 140283726 - 140373392 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04273871_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SCOC-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033939.1 4 - 5 480 178
          GenBank mRNA AK095554.1 - 4 - 5 448 178
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DA487464.1 - 4 - 5 449 Chr. 4 - 140285155 - 140373360 [-] on Build GRCh38 178
          GenBankmRNA DA070842.1 - 4 - 5 491 Chr. 4 - 140285188 - 140373403 [-] on Build GRCh38 178

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream short coiled-coil protein SCOC HRIHFB2072,
          SCOCO,
          UNC-69
          Anti-sense Chr. 4 - 140257286 - 140382556 on NCBI Build GRCh38
          Downstream calmegin CLGN 96.9kb Sense Chr. 4 - 140388453 - 140427661 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0400011969.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC04001588.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P145019 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P281313 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P145019 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 13237 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 2845 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 60554 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P145019 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P281313 - -
          Agilent Human 1B Microarray G4111A A_32_P705811 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 8102872 - -
          Affymetrix Human Gene 1.0 ST Array 8102873 - -
          Affymetrix Human Gene 1.0 ST Array 8102874 - -
          Affymetrix Human Gene 1.1 ST Array 8102871 - -
          Affymetrix HT HG-U133+ PM Array Plate 235869_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 1556736_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR04022125.hg.1 ENST00000512692.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR04022125.hg.1 NR_033939.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04022125.hg.1 TC04001588.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0400011969.hg.1 ENST00000512692.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011969.hg.1 ENST00000512692.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011969.hg.1 ENST00000512692.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011969.hg.1 ENST00000512692.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011969.hg.1 ENST00000512692.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011969.hg.1 ENST00000512692.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011969.hg.1 ENST00000512692.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011969.hg.1 ENST00000512692.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011969.hg.1 ENST00000512692.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011969.hg.1 ENST00000512692.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011969.hg.1 ENST00000608178.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011969.hg.1 ENST00000608178.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011969.hg.1 ENST00000608178.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011969.hg.1 ENST00000608178.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011969.hg.1 ENST00000608178.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011969.hg.1 ENST00000608178.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011969.hg.1 ENST00000608178.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011969.hg.1 ENST00000608178.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011969.hg.1 ENST00000609616.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011969.hg.1 ENST00000609616.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011969.hg.1 ENST00000609616.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011969.hg.1 ENST00000609616.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0400011969.hg.1 NR_033939.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011969.hg.1 NR_033939.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011969.hg.1 NR_033939.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011969.hg.1 NR_033939.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0400011969.hg.1 NR_033939.1 Refseq
          Affymetrix Human Clariom D Assay TC0400011969.hg.1 ENST00000512692.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400011969.hg.1 ENST00000512692.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400011969.hg.1 ENST00000512692.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400011969.hg.1 ENST00000512692.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400011969.hg.1 ENST00000512692.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400011969.hg.1 ENST00000512692.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400011969.hg.1 ENST00000512692.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400011969.hg.1 ENST00000512692.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400011969.hg.1 ENST00000512692.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400011969.hg.1 ENST00000512692.2 Ensembl
          Affymetrix Human Clariom D Assay TC0400011969.hg.1 ENST00000608178.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011969.hg.1 ENST00000608178.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011969.hg.1 ENST00000608178.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011969.hg.1 ENST00000608178.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011969.hg.1 ENST00000608178.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011969.hg.1 ENST00000608178.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011969.hg.1 ENST00000608178.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011969.hg.1 ENST00000608178.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011969.hg.1 ENST00000609616.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011969.hg.1 ENST00000609616.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011969.hg.1 ENST00000609616.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011969.hg.1 ENST00000609616.1 Ensembl
          Affymetrix Human Clariom D Assay TC0400011969.hg.1 NR_033939.1 Refseq
          Affymetrix Human Clariom D Assay TC0400011969.hg.1 NR_033939.1 Refseq
          Affymetrix Human Clariom D Assay TC0400011969.hg.1 NR_033939.1 Refseq
          Affymetrix Human Clariom D Assay TC0400011969.hg.1 NR_033939.1 Refseq
          Affymetrix Human Clariom D Assay TC0400011969.hg.1 NR_033939.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400182077.hg.1 ENST00000512692.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400182077.hg.1 ENST00000512692.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400182077.hg.1 NR_033939.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400182077.hg.1 TC0400011969.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400182077.hg.1 ENST00000512692.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400182077.hg.1 ENST00000512692.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400182077.hg.1 NR_033939.1 Refseq
          Human Genome U133 Plus 2.0 Array 235869_at - -
          Human Genome U133 Plus 2.0 Array 1556736_PM_at - -
          Human Genome U133 Plus 2.0 Array 235869_PM_at - -
          Human Genome U133 Plus 2.0 Array 1556736_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556736_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235869_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235869_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556736_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235869_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235869_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235869_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556736_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235869_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235869_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556736_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556736_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556736_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556736_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556736_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235869_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235869_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235869_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556736_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556736_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556736_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 235869_at4 - -
          Human Genome U95 Set 47380_PM_at - -
          Human Genome U95 Set 47380_at - -
          Human Genome U133 Set 235869_at - -
          Human Genome U133 Set 235869_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A024781 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A024781 - -
          Array Name Probe IDs Targets Target Type
          Illumina human-6 v2.0 expression beadchip ILMN_1841253 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 3450438 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_73263 - -
          Illumina HumanWG-6_V2_0_R2 ILMN_1841253 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0