Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04273907_g1
          See other RNF144A-AS1 GE Assays ›
          Gene Symbol:
          RNF144A-AS1
          Gene Name:
          RNF144A antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 6912276 - 6918682 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04273907_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RNF144A-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033997.1 4 - 5 959 91
          GenBank mRNA AK090737.1 - 2 - 3 248 91
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DA069483.1 - 2 - 3 248 Chr. 2 - 6913341 - 6915912 [-] on Build GRCh38 91
          GenBankmRNA BX376453.2 - 3 - 4 484 Chr. 2 - 6913390 - 6917004 [-] on Build GRCh38 91

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream radical S-adenosyl methionine domain containing 2 RSAD2 2510004L01Rik,
          cig33,
          cig5,
          vig1
          -14.0kb Anti-sense Chr. 2 - 6865733 - 6898239 on NCBI Build GRCh38
          Downstream ring finger protein 144A RNF144A RNF144,
          UBCE7IP4
          3.0kb Anti-sense Chr. 2 - 6917392 - 7076886 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200011627.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02001544.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human CGH Microarray 4x180K 18781 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 48559 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3519424 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 53660 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3519424 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P15542663 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P15542663 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 160539 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 88373 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15542663 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P13094964 - -
          Agilent Human 1B Microarray G4111A A_32_P398877 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P15542663 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1556798_PM_a_at - -
          Affymetrix HT HG-U133+ PM Array Plate 1559145_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 1556797_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR02024976.hg.1 ENST00000418970.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02024976.hg.1 NR_033997.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02024976.hg.1 TC02001544.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR02024975.hg.1 ENST00000418970.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR02024975.hg.1 NR_033997.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02024975.hg.1 TC02001544.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0200184465.hg.1 ENST00000418970.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200184465.hg.1 ENST00000418970.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200184465.hg.1 ENST00000426122.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200184465.hg.1 NR_033997.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200184465.hg.1 TC0200011627.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200184465.hg.1 ENST00000418970.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200184465.hg.1 ENST00000418970.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200184465.hg.1 ENST00000426122.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200184465.hg.1 NR_033997.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200184461.hg.1 ENST00000418970.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200184461.hg.1 ENST00000418970.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200184461.hg.1 NR_033997.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200184461.hg.1 TC0200011627.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200184461.hg.1 ENST00000418970.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200184461.hg.1 ENST00000418970.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200184461.hg.1 NR_033997.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200184463.hg.1 ENST00000418970.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200184463.hg.1 ENST00000418970.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200184463.hg.1 NR_033997.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200184463.hg.1 TC0200011627.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200184463.hg.1 ENST00000418970.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200184463.hg.1 ENST00000418970.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200184463.hg.1 NR_033997.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200184462.hg.1 ENST00000418970.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200184462.hg.1 ENST00000418970.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200184462.hg.1 NR_033997.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200184462.hg.1 TC0200011627.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200184462.hg.1 ENST00000418970.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200184462.hg.1 ENST00000418970.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200184462.hg.1 NR_033997.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200184464.hg.1 ENST00000418970.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200184464.hg.1 ENST00000418970.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200184464.hg.1 ENST00000426122.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200184464.hg.1 NR_033997.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200184464.hg.1 TC0200011627.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200184464.hg.1 ENST00000418970.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200184464.hg.1 ENST00000418970.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200184464.hg.1 ENST00000426122.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0200184464.hg.1 NR_033997.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200184460.hg.1 ENST00000418970.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200184460.hg.1 ENST00000418970.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200184460.hg.1 NR_033997.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200184460.hg.1 TC0200011627.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200184460.hg.1 ENST00000418970.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200184460.hg.1 ENST00000418970.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0200184460.hg.1 NR_033997.1 Refseq
          Human Genome U133 Plus 2.0 Array 1556798_PM_a_at - -
          Human Genome U133 Plus 2.0 Array 1559145_at - -
          Human Genome U133 Plus 2.0 Array 1556798_a_at - -
          Human Genome U133 Plus 2.0 Array 1556797_at - -
          Human Genome U133 Plus 2.0 Array 1556797_PM_at - -
          Human Genome U133 Plus 2.0 Array 1559145_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556797_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559145_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559145_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559145_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556797_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556798_a_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556798_a_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556798_a_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559145_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559145_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559145_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559145_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556798_a_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559145_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556798_a_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556797_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556798_a_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556797_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559145_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556797_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556797_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556797_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556797_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556798_a_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556798_a_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556797_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556797_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556798_a_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559145_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556798_a_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559145_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556797_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556798_a_at7 - -
          Human Genome U95 Set 79051_r_at - -
          Human Genome U95 Set 79051_PM_r_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A013247 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A013247 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0