Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04274232_s1
          See other MGAT4EP GE Assays ›
          Gene Symbol:
          MGAT4EP
          Gene Name:
          MGAT4 family member E, pseudogene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 202820257 - 202827225 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04274232_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MGAT4EP
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038135.2 3 1818 73
          GenBank mRNA BC032956.1 - 3 1815 4827085 73
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DB518250.1 - 1 280 Chr. 1 - 202825945 - 202826371 [+] on Build GRCh38 73
          GenBankmRNA HY214333.1 - 1 Chr. 1 - 202825941 - 202826371 [-] on Build GRCh38 73
          GenBankmRNA DB527564.1 - 1 306 Chr. 1 - 202825919 - 202826371 [+] on Build GRCh38 73

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream prostate cancer associated transcript 6 (non-protein coding) PCAT6 KDM5B-AS1,
          KDM5BAS1,
          PCAN-R1,
          ncRNA-a2,
          onco-lncRNA-96
          -13.3kb Sense Chr. 1 - 202810946 - 202811913 on NCBI Build GRCh38
          Downstream actin pseudogene LOC148709 34.5kb Sense Chr. 1 - 202861754 - 202875241 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100011236.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01001679.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00734556 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20795776 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20795770 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15163965 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15163961 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00734564 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00734551 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00734563 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20795784 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20795755 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20795786 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00734540 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20795759 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20795766 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00734559 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20795760 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00734560 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00734561 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15163966 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00734536 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00734565 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15163963 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00734543 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00734550 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00734557 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00734537 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00734552 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20795753 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 34105 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7908866 - -
          Affymetrix Human Gene 1.0 ST Array 7908864 - -
          Affymetrix Human Gene 1.0 ST Array 7908865 - -
          Affymetrix HT HG-U133+ PM Array Plate 1570023_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0100011236.hg.1 ENST00000330493.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011236.hg.1 ENST00000330493.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011236.hg.1 ENST00000330493.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011236.hg.1 ENST00000330493.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011236.hg.1 ENST00000330493.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011236.hg.1 ENST00000330493.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011236.hg.1 ENST00000330493.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011236.hg.1 ENST00000549576.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100011236.hg.1 NR_038135.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011236.hg.1 NR_038135.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011236.hg.1 NR_038135.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011236.hg.1 NR_038135.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011236.hg.1 NR_038135.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011236.hg.1 NR_038135.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100011236.hg.1 NR_038135.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01026054.hg.1 ENST00000330493.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01026054.hg.1 ENST00000549576.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01026054.hg.1 NR_038135.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01026054.hg.1 TC01001679.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100182465.hg.1 ENST00000330493.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100182465.hg.1 NR_038135.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100182465.hg.1 TC0100011236.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100182465.hg.1 ENST00000330493.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100182465.hg.1 NR_038135.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100182464.hg.1 ENST00000330493.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100182464.hg.1 NR_038135.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100182464.hg.1 TC0100011236.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100182464.hg.1 ENST00000330493.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100182464.hg.1 NR_038135.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100182468.hg.1 ENST00000330493.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100182468.hg.1 NR_038135.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100182468.hg.1 TC0100011236.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100182468.hg.1 ENST00000330493.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100182468.hg.1 NR_038135.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100182463.hg.1 ENST00000330493.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100182463.hg.1 NR_038135.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100182463.hg.1 TC0100011236.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100182463.hg.1 ENST00000330493.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100182463.hg.1 NR_038135.2 Refseq
          Affymetrix Human Clariom D Assay TC0100011236.hg.1 ENST00000330493.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100011236.hg.1 ENST00000330493.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100011236.hg.1 ENST00000330493.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100011236.hg.1 ENST00000330493.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100011236.hg.1 ENST00000330493.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100011236.hg.1 ENST00000330493.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100011236.hg.1 ENST00000330493.6 Ensembl
          Affymetrix Human Clariom D Assay TC0100011236.hg.1 ENST00000549576.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100011236.hg.1 NR_038135.2 Refseq
          Affymetrix Human Clariom D Assay TC0100011236.hg.1 NR_038135.2 Refseq
          Affymetrix Human Clariom D Assay TC0100011236.hg.1 NR_038135.2 Refseq
          Affymetrix Human Clariom D Assay TC0100011236.hg.1 NR_038135.2 Refseq
          Affymetrix Human Clariom D Assay TC0100011236.hg.1 NR_038135.2 Refseq
          Affymetrix Human Clariom D Assay TC0100011236.hg.1 NR_038135.2 Refseq
          Affymetrix Human Clariom D Assay TC0100011236.hg.1 NR_038135.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100182466.hg.1 ENST00000330493.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100182466.hg.1 NR_038135.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100182466.hg.1 TC0100011236.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100182466.hg.1 ENST00000330493.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100182466.hg.1 NR_038135.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100182467.hg.1 ENST00000330493.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100182467.hg.1 ENST00000549576.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100182467.hg.1 NR_038135.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100182467.hg.1 TC0100011236.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100182467.hg.1 ENST00000330493.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100182467.hg.1 ENST00000549576.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0100182467.hg.1 NR_038135.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100182462.hg.1 ENST00000330493.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100182462.hg.1 NR_038135.2 Refseq
          Affymetrix Human Clariom D Assay PSR0100182462.hg.1 TC0100011236.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100182462.hg.1 ENST00000330493.6 Ensembl
          Affymetrix Human Clariom D Assay PSR0100182462.hg.1 NR_038135.2 Refseq
          Human Genome U133 Plus 2.0 Array 1570023_at - -
          Human Genome U133 Plus 2.0 Array 1570023_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1570023_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1570023_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1570023_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1570023_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1570023_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1570023_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1570023_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1570023_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1570023_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1570023_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1570023_at5 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3208464 BC032956.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3208464 NR_038135.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-f85dfd666-px2hc:80/100.66.130.62:80.
          git-commit: 2ca7b3ab9a2195be9b417413331e4e963d93e17c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0