Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009363.hg.1 |
NR_038210.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009363.hg.1 |
TC08000692.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008656.hg.1 |
ENST00000625758.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008656.hg.1 |
ENST00000625758.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008656.hg.1 |
ENST00000625758.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008656.hg.1 |
ENST00000625758.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008656.hg.1 |
ENST00000625758.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008656.hg.1 |
ENST00000629661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008656.hg.1 |
ENST00000629661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008656.hg.1 |
ENST00000629661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008656.hg.1 |
ENST00000629661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008656.hg.1 |
ENST00000629661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008656.hg.1 |
NR_038210.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008656.hg.1 |
NR_038210.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008656.hg.1 |
NR_038210.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008656.hg.1 |
NR_038210.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008656.hg.1 |
NR_038210.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008656.hg.1 |
ENST00000625758.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008656.hg.1 |
ENST00000625758.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008656.hg.1 |
ENST00000625758.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008656.hg.1 |
ENST00000625758.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008656.hg.1 |
ENST00000625758.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008656.hg.1 |
ENST00000629661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008656.hg.1 |
ENST00000629661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008656.hg.1 |
ENST00000629661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008656.hg.1 |
ENST00000629661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008656.hg.1 |
ENST00000629661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008656.hg.1 |
NR_038210.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008656.hg.1 |
NR_038210.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008656.hg.1 |
NR_038210.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008656.hg.1 |
NR_038210.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008656.hg.1 |
NR_038210.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159337.hg.1 |
ENST00000625758.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159337.hg.1 |
ENST00000629661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159337.hg.1 |
NR_038210.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159337.hg.1 |
TC0800008656.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800159337.hg.1 |
ENST00000625758.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159337.hg.1 |
ENST00000629661.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159337.hg.1 |
NR_038210.1 |
Refseq |