Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04274297_m1
          See other LOC100506844 GE Assays ›
          Gene Symbol:
          LOC100506844
          Gene Name:
          uncharacterized LOC100506844
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 57931449 - 57936164 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04274297_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100506844
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038269.1 2 - 3 409 99
          GenBank mRNA BU621729.1 - 1 - 2 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200010988.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12001640.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Recommended Assay Homo sapiens Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379820 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379817 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379815 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28815278 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08379818 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P618928 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR12022195.hg.1 ENST00000547492.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12022195.hg.1 NR_038269.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12022195.hg.1 TC12001640.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR12022191.hg.1 NR_038269.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12022191.hg.1 TC12001640.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR12022192.hg.1 ENST00000547492.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12022192.hg.1 NR_038269.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12022192.hg.1 TC12001640.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR12022196.hg.1 NR_038269.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12022196.hg.1 TC12001640.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR12022193.hg.1 ENST00000547492.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12022193.hg.1 NR_038269.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12022193.hg.1 TC12001640.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1200010988.hg.1 ENST00000546580.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010988.hg.1 ENST00000546580.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010988.hg.1 ENST00000546580.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010988.hg.1 ENST00000546580.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010988.hg.1 ENST00000547492.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010988.hg.1 ENST00000547492.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010988.hg.1 ENST00000547492.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010988.hg.1 ENST00000547492.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010988.hg.1 ENST00000551421.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010988.hg.1 ENST00000551421.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010988.hg.1 ENST00000553102.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010988.hg.1 ENST00000553102.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010988.hg.1 ENST00000553102.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200010988.hg.1 NR_038269.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010988.hg.1 NR_038269.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010988.hg.1 NR_038269.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010988.hg.1 NR_038269.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200010988.hg.1 NR_038269.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200185665.hg.1 ENST00000546580.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200185665.hg.1 ENST00000547492.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200185665.hg.1 ENST00000551421.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200185665.hg.1 NR_038269.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200185665.hg.1 TC1200010988.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200185665.hg.1 NR_038269.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200185665.hg.1 ENST00000546580.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200185665.hg.1 ENST00000547492.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200185665.hg.1 ENST00000551421.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200185668.hg.1 ENST00000546580.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200185668.hg.1 ENST00000547492.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200185668.hg.1 ENST00000553102.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200185668.hg.1 NR_038269.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200185668.hg.1 TC1200010988.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200185668.hg.1 ENST00000546580.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200185668.hg.1 ENST00000547492.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200185668.hg.1 ENST00000553102.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200185668.hg.1 NR_038269.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200185658.hg.1 NR_038269.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200185658.hg.1 TC1200010988.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200185658.hg.1 NR_038269.1 Refseq
          Affymetrix Human Clariom D Assay TC1200010988.hg.1 ENST00000546580.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010988.hg.1 ENST00000546580.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010988.hg.1 ENST00000546580.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010988.hg.1 ENST00000546580.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010988.hg.1 ENST00000547492.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010988.hg.1 ENST00000547492.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010988.hg.1 ENST00000547492.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010988.hg.1 ENST00000547492.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010988.hg.1 ENST00000551421.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010988.hg.1 ENST00000551421.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010988.hg.1 ENST00000553102.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010988.hg.1 ENST00000553102.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010988.hg.1 ENST00000553102.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200010988.hg.1 NR_038269.1 Refseq
          Affymetrix Human Clariom D Assay TC1200010988.hg.1 NR_038269.1 Refseq
          Affymetrix Human Clariom D Assay TC1200010988.hg.1 NR_038269.1 Refseq
          Affymetrix Human Clariom D Assay TC1200010988.hg.1 NR_038269.1 Refseq
          Affymetrix Human Clariom D Assay TC1200010988.hg.1 NR_038269.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200185672.hg.1 ENST00000546580.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200185672.hg.1 ENST00000547492.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200185672.hg.1 ENST00000551421.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200185672.hg.1 ENST00000553102.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200185672.hg.1 NR_038269.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200185672.hg.1 TC1200010988.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200185672.hg.1 ENST00000546580.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200185672.hg.1 ENST00000547492.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200185672.hg.1 ENST00000551421.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200185672.hg.1 ENST00000553102.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200185672.hg.1 NR_038269.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200185677.hg.1 ENST00000546580.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200185677.hg.1 NR_038269.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200185677.hg.1 TC1200010988.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200185677.hg.1 ENST00000546580.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200185677.hg.1 NR_038269.1 Refseq
          Human Genome U133 Plus 2.0 Array 1559492_at - -
          Human Genome U133 Plus 2.0 Array 1559492_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559492_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559492_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559492_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559492_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559492_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559492_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559492_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559492_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559492_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559492_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1559492_at8 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A047742 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A047742 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0