Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04274311_m1
          See other DLG1-AS1 GE Assays ›
          Gene Symbol:
          DLG1-AS1
          Gene Name:
          DLG1 antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 197298247 - 197303751 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04274311_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: DLG1-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038289.1 2 - 3 479 77
          GenBank mRNA BC039329.1 - 2 - 3 474 77
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA HY027083.1 - 3 - 4 398 Chr. 3 - 197298131 - 197302605 [+] on Build GRCh38 77
          GenBankmRNA BI460168.1 - 2 - 3 478 Chr. 3 - 197298247 - 197302698 [+] on Build GRCh38 77
          GenBankmRNA DB462487.1 - 2 - 3 244 Chr. 3 - 197298252 - 197302781 [+] on Build GRCh38 77

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream microRNA 4797 MIR4797 -5.0kb Anti-sense Chr. 3 - 197293878 - 197293948 on NCBI Build GRCh38
          Downstream uncharacterized LOC100129516 LOC100129516 202.4kb Anti-sense Chr. 3 - 197505262 - 197506747 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0300010050.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC03001106.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02882329 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23024865 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23024870 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23024866 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02882341 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23024868 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23024861 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23024857 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23024860 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23024862 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P23024864 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15527944 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02882337 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02882333 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02882326 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02882338 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P02882323 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 445885 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 45122 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 216940 AK057540.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 216940 AK122927.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 216940 AY327404.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 216940 NM_144625.3 refseq_mrna_human_NM
          Agilent SurePrint G3 Human CNV Microarray 2x400K 310539 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 288230 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 21360 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 162633 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P16561602 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P16561609 - -
          Agilent Human 1B Microarray G4111A A_32_P100815 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P16561602 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0300010050.hg.1 ENST00000414529.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010050.hg.1 ENST00000414529.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010050.hg.1 ENST00000414529.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010050.hg.1 ENST00000414529.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010050.hg.1 ENST00000414529.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010050.hg.1 ENST00000414529.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010050.hg.1 ENST00000430666.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010050.hg.1 ENST00000430666.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010050.hg.1 ENST00000430666.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010050.hg.1 ENST00000430666.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010050.hg.1 ENST00000430666.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0300010050.hg.1 NR_038289.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300010050.hg.1 NR_038289.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300010050.hg.1 NR_038289.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300010050.hg.1 NR_038289.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0300010050.hg.1 NR_038289.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03019079.hg.1 ENST00000430666.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03019079.hg.1 NR_038289.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03019079.hg.1 TC03001106.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03019076.hg.1 ENST00000430666.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03019076.hg.1 NR_038289.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03019076.hg.1 TC03001106.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03019078.hg.1 ENST00000430666.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03019078.hg.1 NR_038289.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03019078.hg.1 TC03001106.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03019075.hg.1 ENST00000430666.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03019075.hg.1 NR_038289.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03019075.hg.1 TC03001106.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR03019077.hg.1 ENST00000430666.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR03019077.hg.1 NR_038289.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR03019077.hg.1 TC03001106.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0300173178.hg.1 ENST00000414529.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173178.hg.1 ENST00000414529.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173178.hg.1 ENST00000430666.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173178.hg.1 NR_038289.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300173178.hg.1 TC0300010050.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300173178.hg.1 NR_038289.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300173178.hg.1 ENST00000414529.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173178.hg.1 ENST00000414529.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173178.hg.1 ENST00000430666.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173177.hg.1 ENST00000414529.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173177.hg.1 ENST00000414529.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173177.hg.1 ENST00000430666.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173177.hg.1 NR_038289.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300173177.hg.1 TC0300010050.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300173177.hg.1 ENST00000414529.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173177.hg.1 ENST00000414529.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173177.hg.1 ENST00000430666.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173177.hg.1 NR_038289.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300173176.hg.1 ENST00000430666.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173176.hg.1 NR_038289.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300173176.hg.1 TC0300010050.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300173176.hg.1 ENST00000430666.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173176.hg.1 NR_038289.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300173175.hg.1 ENST00000414529.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173175.hg.1 ENST00000414529.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173175.hg.1 ENST00000430666.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173175.hg.1 NR_038289.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300173175.hg.1 TC0300010050.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300173175.hg.1 ENST00000414529.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173175.hg.1 ENST00000414529.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173175.hg.1 ENST00000430666.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173175.hg.1 NR_038289.1 Refseq
          Affymetrix Human Clariom D Assay TC0300010050.hg.1 ENST00000414529.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300010050.hg.1 ENST00000414529.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300010050.hg.1 ENST00000414529.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300010050.hg.1 ENST00000414529.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300010050.hg.1 ENST00000414529.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300010050.hg.1 ENST00000414529.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300010050.hg.1 ENST00000430666.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300010050.hg.1 ENST00000430666.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300010050.hg.1 ENST00000430666.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300010050.hg.1 ENST00000430666.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300010050.hg.1 ENST00000430666.1 Ensembl
          Affymetrix Human Clariom D Assay TC0300010050.hg.1 NR_038289.1 Refseq
          Affymetrix Human Clariom D Assay TC0300010050.hg.1 NR_038289.1 Refseq
          Affymetrix Human Clariom D Assay TC0300010050.hg.1 NR_038289.1 Refseq
          Affymetrix Human Clariom D Assay TC0300010050.hg.1 NR_038289.1 Refseq
          Affymetrix Human Clariom D Assay TC0300010050.hg.1 NR_038289.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300173179.hg.1 ENST00000430666.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173179.hg.1 NR_038289.1 Refseq
          Affymetrix Human Clariom D Assay PSR0300173179.hg.1 TC0300010050.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0300173179.hg.1 ENST00000430666.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0300173179.hg.1 NR_038289.1 Refseq
          Human Genome U133 Plus 2.0 Array 1556518_at - -
          Human Genome U133 Plus 2.0 Array 1556518_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556518_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556518_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556518_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556518_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556518_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556518_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556518_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556518_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556518_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556518_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1556518_at6 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A023784 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A023784 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0