Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04274368_m1
          See other LOC643770 GE Assays ›
          Gene Symbol:
          LOC643770
          Gene Name:
          uncharacterized LOC643770
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 98485544 - 98503855 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04274368_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC643770
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038383.1 3 - 4 176 73
          GenBank mRNA AK126817.1 - 3 - 4 176 73
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AI654200.1 - 1 - 2 Chr. 12 - 98486974 - 98503787 [+] on Build GRCh38 73
          GenBankmRNA AI792049.1 - 3 - 4 142 Chr. 12 - 98486986 - 98503820 [-] on Build GRCh38 73
          GenBankmRNA BX111292.1 - 3 - 4 141 Chr. 12 - 98486978 - 98503819 [-] on Build GRCh38 73
          GenBankmRNA AA995901.1 - 1 - 2 Chr. 12 - 98486971 - 98493488 [+] on Build GRCh38 73
          GenBankmRNA AI820655.1 - 1 - 2 Chr. 12 - 98486971 - 98503787 [+] on Build GRCh38 73

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream solute carrier family 9 member 7 pseudogene 1 SLC9A7P1 -28.4kb Sense Chr. 12 - 98453840 - 98457145 on NCBI Build GRCh38
          Downstream TMPO antisense RNA 1 TMPO-AS1 19.5kb Sense Chr. 12 - 98512973 - 98516226 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200011646.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12001858.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28995710 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08562344 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08562280 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28995711 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28995668 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08562337 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28995684 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28995713 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28995629 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08562279 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28995626 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28995649 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08562281 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08562335 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08562312 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08562286 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08562313 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08562330 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08562338 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08562300 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28995715 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28995634 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28995653 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28995636 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08562299 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28995672 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28995624 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08562332 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08562307 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28995659 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08562304 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28995656 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28995687 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08562292 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08562321 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28995655 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08562293 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08562331 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08562346 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08562298 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28995639 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08562271 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08562306 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28995660 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28995637 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28995664 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28995709 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28995628 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08562277 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16584312 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28995712 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08562327 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28995625 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28995683 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08562334 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28995686 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08562310 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16584310 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28995696 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28995692 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P28995651 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 156404 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 719124 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 564768 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 856980 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 610927 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 880119 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 373596 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 209952 CR590757.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 209952 XM_001129364.1 refseq_mrna_human_XM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 209952 XM_377761.4 refseq_mrna_human_XM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 209952 XM_928270.2 refseq_mrna_human_XM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 518907 BC032648.1 genbank_mrna_human
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3309414 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 28366 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P19652659 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P56911032 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3309414 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3309414 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P19652659 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P19652667 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P19652659 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P39786124 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P19652673 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P19652667 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P39786111 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P19652653 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P19652667 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P39786111 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P19652659 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR12024335.hg.1 ENST00000501499.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12024335.hg.1 ENST00000549004.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12024335.hg.1 NR_038383.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12024335.hg.1 TC12001858.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR12024338.hg.1 ENST00000501499.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12024338.hg.1 NR_038383.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12024338.hg.1 TC12001858.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR12024334.hg.1 ENST00000501499.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12024334.hg.1 NR_038383.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12024334.hg.1 TC12001858.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1200011646.hg.1 ENST00000501499.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011646.hg.1 ENST00000501499.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011646.hg.1 ENST00000501499.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011646.hg.1 ENST00000501499.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011646.hg.1 ENST00000501499.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011646.hg.1 ENST00000501499.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011646.hg.1 ENST00000501499.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011646.hg.1 ENST00000549004.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011646.hg.1 ENST00000549004.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011646.hg.1 ENST00000549004.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011646.hg.1 NR_038383.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011646.hg.1 NR_038383.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011646.hg.1 NR_038383.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011646.hg.1 NR_038383.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011646.hg.1 NR_038383.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011646.hg.1 NR_038383.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011646.hg.1 NR_038383.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12024337.hg.1 ENST00000501499.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12024337.hg.1 NR_038383.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12024337.hg.1 TC12001858.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR12024339.hg.1 ENST00000501499.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12024339.hg.1 NR_038383.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12024339.hg.1 TC12001858.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1200190138.hg.1 ENST00000501499.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200190138.hg.1 NR_038383.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200190138.hg.1 TC1200011646.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200190138.hg.1 ENST00000501499.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200190138.hg.1 NR_038383.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200190142.hg.1 ENST00000501499.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200190142.hg.1 ENST00000549004.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200190142.hg.1 NR_038383.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200190142.hg.1 TC1200011646.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200190142.hg.1 ENST00000501499.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200190142.hg.1 ENST00000549004.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200190142.hg.1 NR_038383.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200190137.hg.1 ENST00000501499.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200190137.hg.1 NR_038383.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200190137.hg.1 TC1200011646.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200190137.hg.1 ENST00000501499.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200190137.hg.1 NR_038383.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200190141.hg.1 ENST00000501499.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200190141.hg.1 ENST00000549004.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200190141.hg.1 NR_038383.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200190141.hg.1 TC1200011646.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200190141.hg.1 ENST00000501499.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200190141.hg.1 ENST00000549004.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200190141.hg.1 NR_038383.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200190150.hg.1 ENST00000501499.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200190150.hg.1 NR_038383.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200190150.hg.1 TC1200011646.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200190150.hg.1 ENST00000501499.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200190150.hg.1 NR_038383.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200190147.hg.1 ENST00000501499.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200190147.hg.1 NR_038383.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200190147.hg.1 TC1200011646.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200190147.hg.1 ENST00000501499.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200190147.hg.1 NR_038383.1 Refseq
          Affymetrix Human Clariom D Assay TC1200011646.hg.1 ENST00000501499.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011646.hg.1 ENST00000501499.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011646.hg.1 ENST00000501499.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011646.hg.1 ENST00000501499.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011646.hg.1 ENST00000501499.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011646.hg.1 ENST00000501499.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011646.hg.1 ENST00000501499.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011646.hg.1 ENST00000549004.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011646.hg.1 ENST00000549004.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011646.hg.1 ENST00000549004.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011646.hg.1 NR_038383.1 Refseq
          Affymetrix Human Clariom D Assay TC1200011646.hg.1 NR_038383.1 Refseq
          Affymetrix Human Clariom D Assay TC1200011646.hg.1 NR_038383.1 Refseq
          Affymetrix Human Clariom D Assay TC1200011646.hg.1 NR_038383.1 Refseq
          Affymetrix Human Clariom D Assay TC1200011646.hg.1 NR_038383.1 Refseq
          Affymetrix Human Clariom D Assay TC1200011646.hg.1 NR_038383.1 Refseq
          Affymetrix Human Clariom D Assay TC1200011646.hg.1 NR_038383.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200190149.hg.1 ENST00000501499.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200190149.hg.1 NR_038383.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200190149.hg.1 TC1200011646.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200190149.hg.1 ENST00000501499.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200190149.hg.1 NR_038383.1 Refseq
          Human Genome U95 Set 77625_at - -
          Human Genome U95 Set 77625_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A047923 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A047923 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1752978 NR_038383.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1752978 AK126817.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1752978 NR_038383.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1752978 AK126817.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1752978 NR_038383.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1752978 AK126817.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1752978 NR_038383.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1752978 AK126817.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1752978 NR_038383.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1752978 AK126817.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0