Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04274384_m1
          See other STAU2-AS1 GE Assays ›
          Gene Symbol:
          STAU2-AS1
          Gene Name:
          STAU2 antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 73420074 - 73441518 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04274384_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: STAU2-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038406.1 1 - 2 161 88
          GenBank mRNA AK056720.1 - 1 - 2 161 88
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DB224181.1 - 1 - 2 233 Chr. 8 - 73420004 - 73439237 [+] on Build GRCh38 88
          GenBankmRNA DA835077.1 - 1 - 2 161 Chr. 8 - 73420075 - 73439998 [+] on Build GRCh38 88

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RDH10 antisense RNA 1 RDH10-AS1 -63.6kb Anti-sense Chr. 8 - 73310906 - 73356461 on NCBI Build GRCh38
          Downstream ubiquitin conjugating enzyme E2 W (putative) UBE2W UBC-16,
          UBC16
          341.0kb Anti-sense Chr. 8 - 73780097 - 73878910 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0800007959.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC08000484.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214769 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214908 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26497954 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16106441 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16106440 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26497846 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214805 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214782 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214817 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26497864 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214821 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26497967 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26497843 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214819 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214773 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214910 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214903 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16106404 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26497862 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214824 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26497865 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16106400 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26497828 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214780 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214795 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26497817 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214767 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26497855 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214785 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214801 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214760 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26497840 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214915 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214764 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26497845 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214790 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16106412 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214766 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214789 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16106414 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16106413 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26497937 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16106416 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26497834 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26497849 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26497945 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214919 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214899 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214771 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214792 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26497802 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214809 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214902 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214901 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214778 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26497944 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26497856 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26497955 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16106443 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214794 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26497847 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214909 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214898 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214787 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214890 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26497958 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16106411 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214770 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16106444 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26497833 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26497957 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214772 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214761 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214894 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214912 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26497936 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26497861 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26497829 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214781 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214799 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06214793 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 33709 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3355408 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P18353084 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P18353084 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P01943033 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 169517 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 76368 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01943048 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18353084 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P32289057 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18353077 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18353069 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01943033 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01943034 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18353106 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18353096 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1561391_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR08006682.hg.1 ENST00000517604.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08006682.hg.1 NR_038406.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08006682.hg.1 TC08000484.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0800007959.hg.1 ENST00000517604.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007959.hg.1 ENST00000517604.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007959.hg.1 ENST00000517604.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007959.hg.1 ENST00000522703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007959.hg.1 ENST00000522703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007959.hg.1 ENST00000522703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007959.hg.1 ENST00000522703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800007959.hg.1 NR_038406.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800007959.hg.1 NR_038406.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800007959.hg.1 NR_038406.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08006680.hg.1 ENST00000517604.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08006680.hg.1 ENST00000522703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08006680.hg.1 NR_038406.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08006680.hg.1 TC08000484.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR08006679.hg.1 ENST00000517604.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08006679.hg.1 ENST00000522703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08006679.hg.1 NR_038406.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08006679.hg.1 TC08000484.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0800007959.hg.1 ENST00000517604.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007959.hg.1 ENST00000517604.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007959.hg.1 ENST00000517604.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007959.hg.1 ENST00000522703.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007959.hg.1 ENST00000522703.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007959.hg.1 ENST00000522703.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007959.hg.1 ENST00000522703.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800007959.hg.1 NR_038406.1 Refseq
          Affymetrix Human Clariom D Assay TC0800007959.hg.1 NR_038406.1 Refseq
          Affymetrix Human Clariom D Assay TC0800007959.hg.1 NR_038406.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800154871.hg.1 ENST00000517604.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800154871.hg.1 ENST00000522703.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800154871.hg.1 NR_038406.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800154871.hg.1 TC0800007959.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800154871.hg.1 NR_038406.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800154871.hg.1 ENST00000517604.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800154871.hg.1 ENST00000522703.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800154874.hg.1 ENST00000517604.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800154874.hg.1 NR_038406.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800154874.hg.1 TC0800007959.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800154874.hg.1 ENST00000517604.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800154874.hg.1 NR_038406.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800154872.hg.1 ENST00000517604.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800154872.hg.1 ENST00000522703.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800154872.hg.1 NR_038406.1 Refseq
          Affymetrix Human Clariom D Assay PSR0800154872.hg.1 TC0800007959.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800154872.hg.1 ENST00000517604.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800154872.hg.1 ENST00000522703.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800154872.hg.1 NR_038406.1 Refseq
          Human Genome U133 Plus 2.0 Array 1561391_PM_at - -
          Human Genome U133 Plus 2.0 Array 1561391_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561391_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561391_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561391_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561391_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561391_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561391_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561391_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561391_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561391_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561391_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1561391_at1 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A036692 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A036692 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3178553 NR_038406.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3178553 AK056720.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3269812 NR_038406.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3269812 AK056720.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0