Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04274458_m1
          See other LINC01289 GE Assays ›
          Gene Symbol:
          LINC01289
          Gene Name:
          long intergenic non-protein coding RNA 1289
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 63769430 - 63785501 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04274458_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC01289
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038875.2 2 - 3 2884 81
          GenBank mRNA BC037345.2 - 2 - 3 2884 81

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC102724612 LOC102724612 -303.0kb Sense Chr. 8 - 63465849 - 63475482 on NCBI Build GRCh38
          Downstream uncharacterized LOC102724623 LOC102724623 71.4kb Anti-sense Chr. 8 - 63856943 - 64368558 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0800012304.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC08000429.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06171123 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454155 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454196 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06171026 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454162 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454198 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454246 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454222 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454237 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454250 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454202 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06171063 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454165 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454194 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454235 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454228 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454209 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454142 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454214 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454157 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454219 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454152 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454199 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454262 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454227 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454187 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454212 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454232 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06171032 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454225 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454236 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454160 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06171053 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454156 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454167 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454240 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454251 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06171070 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454257 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454185 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06171084 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454203 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454229 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454238 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06171153 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454223 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454181 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06171136 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454148 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454221 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454189 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06171079 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454153 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454168 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454138 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06171058 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06171147 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06171064 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06171109 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454245 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06171104 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454206 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454226 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454208 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454247 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06171042 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06171157 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454188 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454233 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454211 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454186 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454159 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454145 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454147 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454146 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454200 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454197 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06171077 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454204 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454158 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454169 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06171117 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454150 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454220 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454244 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06171059 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454230 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454172 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454164 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06171037 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06171028 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454205 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454173 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06171071 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454179 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454231 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06171080 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454151 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06171149 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454234 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06171107 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P26454207 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P06171134 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 969163 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 954174 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 629791 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 335586 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 278073 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 273251 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 157411 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 71371 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 309287 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 92109 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 22141 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 160736 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 29403 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 232117 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3503990 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 37278 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3503990 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3503990 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 15474 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P38422944 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18330289 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18330284 - -
          Agilent Human 1B Microarray G4111A A_32_P713116 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P18330289 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1560337_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR08005961.hg.1 ENST00000519550.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR08005961.hg.1 NR_038875.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR08005961.hg.1 TC08000429.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0800012304.hg.1 ENST00000519550.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012304.hg.1 ENST00000519550.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012304.hg.1 ENST00000519550.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012304.hg.1 ENST00000519550.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012304.hg.1 ENST00000519550.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012304.hg.1 ENST00000519550.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800012304.hg.1 NR_038875.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800012304.hg.1 NR_038875.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800012304.hg.1 NR_038875.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800012304.hg.1 NR_038875.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800012304.hg.1 NR_038875.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800012304.hg.1 NR_038875.2 Refseq
          Affymetrix Human Clariom D Assay TC0800012304.hg.1 ENST00000519550.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800012304.hg.1 ENST00000519550.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800012304.hg.1 ENST00000519550.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800012304.hg.1 ENST00000519550.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800012304.hg.1 ENST00000519550.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800012304.hg.1 ENST00000519550.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800012304.hg.1 NR_038875.2 Refseq
          Affymetrix Human Clariom D Assay TC0800012304.hg.1 NR_038875.2 Refseq
          Affymetrix Human Clariom D Assay TC0800012304.hg.1 NR_038875.2 Refseq
          Affymetrix Human Clariom D Assay TC0800012304.hg.1 NR_038875.2 Refseq
          Affymetrix Human Clariom D Assay TC0800012304.hg.1 NR_038875.2 Refseq
          Affymetrix Human Clariom D Assay TC0800012304.hg.1 NR_038875.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800185381.hg.1 ENST00000519550.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800185381.hg.1 NR_038875.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800185381.hg.1 TC0800012304.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800185381.hg.1 NR_038875.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800185381.hg.1 ENST00000519550.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800185382.hg.1 ENST00000519550.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800185382.hg.1 NR_038875.2 Refseq
          Affymetrix Human Clariom D Assay PSR0800185382.hg.1 TC0800012304.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0800185382.hg.1 ENST00000519550.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0800185382.hg.1 NR_038875.2 Refseq
          Human Genome U133 Plus 2.0 Array 1560337_PM_at - -
          Human Genome U133 Plus 2.0 Array 1560337_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560337_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560337_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560337_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560337_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560337_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560337_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560337_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560337_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560337_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560337_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1560337_at11 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A036653 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A036653 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0