Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04274466_m1
          See other LOC285000 GE Assays ›
          Gene Symbol:
          LOC285000
          Gene Name:
          uncharacterized LOC285000
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 105593097 - 105610559 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04274466_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC285000
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038891.1 3 - 4 646 82
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BF690215.1 - Chr. - - [-] on Build 82
          GenBankmRNA BU195220.1 - 3 - 4 544 Chr. 2 - 105593602 - 105610457 [-] on Build GRCh38 82
          GenBankmRNA BM550785.1 - 2 - 3 322 Chr. 2 - 105593128 - 105601141 [-] on Build GRCh38 82
          GenBankmRNA BF690323.1 - 1 - 2 Chr. 2 - 105593269 - 105601071 [+] on Build GRCh38 82
          GenBankmRNA BG768842.1 - 3 - 4 490 Chr. 2 - 105593583 - 105610399 [-] on Build GRCh38 82
          GenBankmRNA BF690014.1 - 1 - 2 Chr. 2 - 105593275 - 105597228 [+] on Build GRCh38 82
          GenBankmRNA BG769143.1 - 3 - 4 488 Chr. 2 - 105593388 - 105610399 [-] on Build GRCh38 82
          GenBankmRNA BG762757.1 - 3 - 4 488 Chr. 2 - 105593496 - 105610399 [-] on Build GRCh38 82
          GenBankmRNA BG770600.1 - 3 - 4 488 Chr. 2 - 105593589 - 105610399 [-] on Build GRCh38 82

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream four and a half LIM domains 2 FHL2 AAG11,
          DRAL,
          FHL-2,
          SLIM-3,
          SLIM3
          -154.3kb Sense Chr. 2 - 105360826 - 105438773 on NCBI Build GRCh38
          Downstream NCK adaptor protein 2 NCK2 GRB4,
          NCKbeta
          147.7kb Anti-sense Chr. 2 - 105744649 - 105894274 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0200013753.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC02002168.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486616 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383096 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383158 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486581 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486669 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383161 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383272 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383291 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383152 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383091 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486725 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383256 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383238 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383154 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486542 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486593 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383172 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486708 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383138 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486625 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383255 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486545 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486572 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486662 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486697 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383116 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383187 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486628 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383222 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383134 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486555 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383147 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486556 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383127 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486663 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383270 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383164 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486672 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383215 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383126 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383133 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486702 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15295444 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383275 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383137 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383245 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486638 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486618 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383089 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486559 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486582 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383226 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15295438 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486681 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486644 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383174 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486727 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383263 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486684 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383100 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383214 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383128 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486631 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486620 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383196 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383149 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383085 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383204 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383119 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486539 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383159 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383124 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486594 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383229 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486533 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486685 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486712 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486682 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383242 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383084 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486649 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383117 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486643 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383107 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486575 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486695 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383145 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486653 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486611 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486722 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383273 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486667 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486604 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383086 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383205 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383231 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383170 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383262 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383195 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383157 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383114 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486676 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486548 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486724 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383113 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383203 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383136 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486577 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486691 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383240 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383253 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383156 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486733 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486627 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486574 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486534 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486563 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486551 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486715 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383160 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383265 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383190 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486655 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383183 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486738 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383146 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383200 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486729 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383259 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15295434 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383099 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383292 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383218 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486583 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486613 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486584 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383295 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383286 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486686 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383110 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486552 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383108 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383252 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486538 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486536 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486637 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486677 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383139 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486659 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486678 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486544 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486689 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383189 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486674 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486573 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383201 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383258 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486530 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486666 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383140 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383102 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383257 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486736 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P21486600 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383276 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383182 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P01383175 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 157642 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 592959 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 366880 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 685407 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 285328 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 770434 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 60738 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 407297 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3579336 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 27790 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P35797769 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P35797769 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3579336 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3579336 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P35797769 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P35797769 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 125089 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 37454 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P35797769 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P35797777 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15769694 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15769714 - -
          Agilent Human 1B Microarray G4111A A_32_P334790 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P35797769 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 240428_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR02035285.hg.1 NR_038891.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR02035285.hg.1 TC02002168.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0200200856.hg.1 NR_038891.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200200856.hg.1 TC0200013753.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200200856.hg.1 NR_038891.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200200855.hg.1 NR_038891.1 Refseq
          Affymetrix Human Clariom D Assay PSR0200200855.hg.1 TC0200013753.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0200200855.hg.1 NR_038891.1 Refseq
          Human Genome U133 Plus 2.0 Array 240428_at - -
          Human Genome U133 Plus 2.0 Array 240428_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240428_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240428_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240428_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240428_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240428_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240428_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240428_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240428_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240428_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240428_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240428_at4 - -
          Human Genome U133 Set 240428_PM_at - -
          Human Genome U133 Set 240428_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A014144 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A014144 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0