Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04274507_m1
          See other HTR5A-AS1 GE Assays ›
          Gene Symbol:
          HTR5A-AS1
          Gene Name:
          HTR5A antisense RNA 1
          Gene Aliases:
          HTR5AOS
          Chromosome Location:
          Chr.7: 155067067 - 155071557 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04274507_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HTR5A-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038945.1 1 - 2 523 76
          GenBank mRNA BC031272.1 - 1 - 2 497 76
          BC045795.1 - 1 - 2 500 76
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DA276425.1 - 1 - 2 235 Chr. 7 - 155068906 - 155071269 [-] on Build GRCh38 76
          GenBankmRNA DA168347.1 - 1 - 2 152 Chr. 7 - 155068843 - 155071166 [-] on Build GRCh38 76
          GenBankmRNA BM926157.1 - 201 Chr. - - [-] on Build 76
          GenBankmRNA BI603546.1 - 1 - 2 503 Chr. 7 - 155068947 - 155071530 [-] on Build GRCh38 76
          GenBankmRNA DB176520.1 - 1 - 2 217 Chr. 7 - 155068915 - 155071249 [-] on Build GRCh38 76
          GenBankmRNA DB486743.1 - 1 - 2 287 Chr. 7 - 155069104 - 155071320 [-] on Build GRCh38 76
          GenBankmRNA DA176219.1 - 1 - 2 217 Chr. 7 - 155068895 - 155071248 [-] on Build GRCh38 76

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream PAXIP1 antisense RNA 1 (head to head) PAXIP1-AS1 -61.4kb Anti-sense Chr. 7 - 155003433 - 155005703 on NCBI Build GRCh38
          Downstream insulin induced gene 1 INSIG1 CL-6,
          CL6
          226.2kb Anti-sense Chr. 7 - 155297776 - 155310235 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0700013163.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC07002055.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05885710 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05885709 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05885712 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05885711 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05885707 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P05885708 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3375279 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 53704 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3375279 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P01840576 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01840576 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P18175491 - -
          Agilent Human 1B Microarray G4111A A_32_P207270 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR07031533.hg.1 ENST00000395731.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07031533.hg.1 ENST00000493904.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07031533.hg.1 ENST00000543018.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07031533.hg.1 NR_038945.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07031533.hg.1 TC07002055.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0700013163.hg.1 ENST00000395731.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013163.hg.1 ENST00000395731.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013163.hg.1 ENST00000395731.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013163.hg.1 ENST00000395731.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013163.hg.1 ENST00000493904.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013163.hg.1 ENST00000493904.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0700013163.hg.1 NR_038945.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013163.hg.1 NR_038945.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013163.hg.1 NR_038945.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0700013163.hg.1 NR_038945.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07031530.hg.1 ENST00000395731.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07031530.hg.1 ENST00000543018.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07031530.hg.1 NR_038945.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07031530.hg.1 TC07002055.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR07031529.hg.1 ENST00000395731.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR07031529.hg.1 NR_038945.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR07031529.hg.1 TC07002055.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0700194979.hg.1 ENST00000395731.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700194979.hg.1 NR_038945.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700194979.hg.1 TC0700013163.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700194979.hg.1 NR_038945.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700194979.hg.1 ENST00000395731.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013163.hg.1 ENST00000395731.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013163.hg.1 ENST00000395731.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013163.hg.1 ENST00000395731.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013163.hg.1 ENST00000395731.5 Ensembl
          Affymetrix Human Clariom D Assay TC0700013163.hg.1 ENST00000493904.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700013163.hg.1 ENST00000493904.2 Ensembl
          Affymetrix Human Clariom D Assay TC0700013163.hg.1 NR_038945.1 Refseq
          Affymetrix Human Clariom D Assay TC0700013163.hg.1 NR_038945.1 Refseq
          Affymetrix Human Clariom D Assay TC0700013163.hg.1 NR_038945.1 Refseq
          Affymetrix Human Clariom D Assay TC0700013163.hg.1 NR_038945.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700194978.hg.1 ENST00000395731.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700194978.hg.1 NR_038945.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700194978.hg.1 TC0700013163.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700194978.hg.1 ENST00000395731.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700194978.hg.1 NR_038945.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700194983.hg.1 ENST00000395731.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700194983.hg.1 ENST00000493904.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700194983.hg.1 NR_038945.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700194983.hg.1 TC0700013163.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700194983.hg.1 ENST00000395731.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700194983.hg.1 ENST00000493904.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0700194983.hg.1 NR_038945.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700194980.hg.1 ENST00000395731.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700194980.hg.1 NR_038945.1 Refseq
          Affymetrix Human Clariom D Assay PSR0700194980.hg.1 TC0700013163.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0700194980.hg.1 ENST00000395731.5 Ensembl
          Affymetrix Human Clariom D Assay PSR0700194980.hg.1 NR_038945.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A033796 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A033796 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3182017 BC031272.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3182017 BC045795.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3182017 DC407680.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3182017 NR_038945.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3182017 AK303289.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix