Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04274725_m1
          See other VWA8-AS1 GE Assays ›
          Gene Symbol:
          VWA8-AS1
          Gene Name:
          VWA8 antisense RNA 1 (head to head)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 41961169 - 41981565 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04274725_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: VWA8-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_039974.1 3 - 4 1302 85
          GenBank mRNA AK092199.1 - 3 - 4 1302 85
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA HY012147.1 - 3 - 4 364 Chr. 13 - 41955730 - 41980947 [+] on Build GRCh38 85
          GenBankmRNA BX098944.1 - 3 - 4 285 Chr. 13 - 41955808 - 41980977 [+] on Build GRCh38 85

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream von Willebrand factor A domain containing 8 VWA8 KIAA0564 -15.3kb Anti-sense Chr. 13 - 41566825 - 41961111 on NCBI Build GRCh38
          Downstream diacylglycerol kinase eta DGKH DGKeta 58.5kb Sense Chr. 13 - 42040036 - 42256584 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1300006985.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC13000149.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Homo sapiens 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253823 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253859 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08806410 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253833 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253792 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253789 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253843 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253825 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253815 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253861 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253865 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08806379 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253795 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253857 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253870 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253844 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253799 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08806419 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08806372 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253828 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253847 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253818 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08806424 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08806438 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08806361 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253856 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08806391 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253817 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08806376 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253781 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08806421 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253839 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08806422 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08806439 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253863 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253860 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253824 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08806427 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253838 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253813 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253846 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253854 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253812 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253827 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253855 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08806360 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253784 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253829 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253804 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253783 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253805 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08806368 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08806386 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08806378 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08806395 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P16630121 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253845 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253842 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P08806397 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253835 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P29253858 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 31308 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 71135 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 486918 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 582239 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 230142 AK131274.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 230142 BC017966.1 genbank_mrna_human
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 230142 NM_001033723.1 refseq_mrna_human_NM
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 792627 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 402324 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 298441 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 636471 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 72588 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 209956 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 382429 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 27712 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 389921 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 283931 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P19790348 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P19790325 - -
          Agilent Human Genome CGH Microarray 105A (G4412A) A_16_P19790348 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P19790325 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P19790348 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 45074 - -
          Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 95401 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P39928717 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P19790354 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P19790348 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P11716185 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P11714922 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P19790350 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P19790357 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02764031 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02764037 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P02764052 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P19790325 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P39928733 - -
          Agilent Human 1B Microarray G4111A A_32_P851913 - -
          Agilent Human 1B Microarray G4111A A_32_P801787 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P02764031 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P19790357 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P19790325 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P19790348 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_18_P11716185 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR13001725.hg.1 ENST00000501265.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR13001725.hg.1 NR_039974.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR13001725.hg.1 TC13000149.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1300006985.hg.1 ENST00000611103.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006985.hg.1 ENST00000611103.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006985.hg.1 ENST00000611103.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006985.hg.1 ENST00000611103.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006985.hg.1 ENST00000611103.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006985.hg.1 ENST00000611103.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006985.hg.1 ENST00000612345.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006985.hg.1 ENST00000612345.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006985.hg.1 ENST00000612345.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006985.hg.1 ENST00000612345.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300006985.hg.1 NR_039974.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006985.hg.1 NR_039974.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006985.hg.1 NR_039974.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006985.hg.1 NR_039974.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006985.hg.1 NR_039974.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300006985.hg.1 NR_039974.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300147382.hg.1 ENST00000611103.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300147382.hg.1 NR_039974.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300147382.hg.1 TC1300006985.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300147382.hg.1 NR_039974.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300147382.hg.1 ENST00000611103.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300147381.hg.1 ENST00000611103.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300147381.hg.1 ENST00000612345.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1300147381.hg.1 NR_039974.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300147381.hg.1 TC1300006985.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300147381.hg.1 ENST00000611103.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300147381.hg.1 ENST00000612345.4 Ensembl
          Affymetrix Human Clariom D Assay PSR1300147381.hg.1 NR_039974.1 Refseq
          Affymetrix Human Clariom D Assay TC1300006985.hg.1 ENST00000611103.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300006985.hg.1 ENST00000611103.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300006985.hg.1 ENST00000611103.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300006985.hg.1 ENST00000611103.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300006985.hg.1 ENST00000611103.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300006985.hg.1 ENST00000611103.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300006985.hg.1 ENST00000612345.4 Ensembl
          Affymetrix Human Clariom D Assay TC1300006985.hg.1 ENST00000612345.4 Ensembl
          Affymetrix Human Clariom D Assay TC1300006985.hg.1 ENST00000612345.4 Ensembl
          Affymetrix Human Clariom D Assay TC1300006985.hg.1 ENST00000612345.4 Ensembl
          Affymetrix Human Clariom D Assay TC1300006985.hg.1 NR_039974.1 Refseq
          Affymetrix Human Clariom D Assay TC1300006985.hg.1 NR_039974.1 Refseq
          Affymetrix Human Clariom D Assay TC1300006985.hg.1 NR_039974.1 Refseq
          Affymetrix Human Clariom D Assay TC1300006985.hg.1 NR_039974.1 Refseq
          Affymetrix Human Clariom D Assay TC1300006985.hg.1 NR_039974.1 Refseq
          Affymetrix Human Clariom D Assay TC1300006985.hg.1 NR_039974.1 Refseq
          Human Genome U133 Plus 2.0 Array 1557646_at - -
          Human Genome U133 Plus 2.0 Array 1557646_PM_at - -
          Human Genome U133 Plus 2.0 Array 1557647_PM_a_at - -
          Human Genome U133 Plus 2.0 Array 1557647_a_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557647_a_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557647_a_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557646_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557646_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557646_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557646_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557647_a_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557646_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557647_a_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557647_a_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557647_a_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557647_a_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557647_a_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557646_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557646_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557646_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557647_a_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557647_a_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557646_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557646_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557646_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1557647_a_at11 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A050097 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A050097 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 235362 - -
          Array Name Probe IDs Targets Target Type
          Illumina human-6 v2.0 expression beadchip ILMN_1849638 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 6940332 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_78577 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1849638 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1849638 - -
          Illumina HumanWG-6_V2_0_R2 ILMN_1849638 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0