Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04274727_m1
          See other SEC24B-AS1 GE Assays ›
          Gene Symbol:
          SEC24B-AS1
          Gene Name:
          SEC24B antisense RNA 1
          Gene Aliases:
          1/2-SBSRNA4
          Chromosome Location:
          Chr.4: 109429963 - 109433817 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04274727_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SEC24B-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_039978.1 1 - 2 89 72
          GenBank mRNA BC009800.1 - 1 - 2 89 72
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DA570698.1 - 1 - 2 55 Chr. 4 - 109357285 - 109433783 [-] on Build GRCh38 72
          GenBankmRNA BF381619.1 - 86 Chr. - - [-] on Build 72

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream zinc finger CCHC-type containing 23 ZCCHC23 -110.9kb Anti-sense Chr. 4 - 109303035 - 109320852 on NCBI Build GRCh38
          Downstream microRNA 576 MIR576 MIRN576,
          hsa-mir-576
          54.9kb Anti-sense Chr. 4 - 109488698 - 109488795 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0400012954.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC04001463.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human CGH Microarray 4x180K 17844 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 384128 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_24_P744148 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 3812 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P01049737 - -
          Whole Human Genome Oligo Microarray G4112A A_24_P744148 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P01049737 - -
          Agilent Whole Human Genome Microarray 4x44K G4112F A_24_P744148 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01049737 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P01049737 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR04020788.hg.1 NR_039978.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04020788.hg.1 TC04001463.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR04020789.hg.1 NR_039978.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04020789.hg.1 TC04001463.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR04020786.hg.1 NR_039978.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04020786.hg.1 TC04001463.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR04020791.hg.1 NR_039978.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04020791.hg.1 TC04001463.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR04020785.hg.1 NR_039978.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR04020785.hg.1 TC04001463.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0400190868.hg.1 ENST00000499359.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190868.hg.1 NR_039978.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400190868.hg.1 TC0400012954.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400190868.hg.1 NR_039978.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400190868.hg.1 ENST00000499359.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190873.hg.1 ENST00000499359.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190873.hg.1 ENST00000505895.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190873.hg.1 ENST00000505895.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190873.hg.1 ENST00000510971.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190873.hg.1 ENST00000510971.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190873.hg.1 NR_039978.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400190873.hg.1 TC0400012954.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400190873.hg.1 ENST00000499359.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190873.hg.1 ENST00000505895.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190873.hg.1 ENST00000505895.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190873.hg.1 ENST00000510971.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190873.hg.1 ENST00000510971.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190873.hg.1 NR_039978.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400190877.hg.1 ENST00000499359.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190877.hg.1 ENST00000505895.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190877.hg.1 ENST00000505895.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190877.hg.1 ENST00000510971.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190877.hg.1 ENST00000510971.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190877.hg.1 NR_039978.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400190877.hg.1 TC0400012954.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400190877.hg.1 ENST00000499359.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190877.hg.1 ENST00000505895.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190877.hg.1 ENST00000505895.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190877.hg.1 ENST00000510971.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190877.hg.1 ENST00000510971.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190877.hg.1 NR_039978.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400190866.hg.1 ENST00000499359.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190866.hg.1 NR_039978.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400190866.hg.1 TC0400012954.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400190866.hg.1 ENST00000499359.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190866.hg.1 NR_039978.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400190869.hg.1 ENST00000499359.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190869.hg.1 ENST00000510971.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190869.hg.1 ENST00000510971.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190869.hg.1 NR_039978.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400190869.hg.1 TC0400012954.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400190869.hg.1 ENST00000499359.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190869.hg.1 ENST00000510971.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190869.hg.1 ENST00000510971.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190869.hg.1 NR_039978.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400190879.hg.1 ENST00000499359.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190879.hg.1 NR_039978.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400190879.hg.1 TC0400012954.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400190879.hg.1 ENST00000499359.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190879.hg.1 NR_039978.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400190867.hg.1 ENST00000499359.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190867.hg.1 NR_039978.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400190867.hg.1 TC0400012954.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400190867.hg.1 ENST00000499359.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190867.hg.1 NR_039978.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400190871.hg.1 ENST00000499359.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190871.hg.1 ENST00000510971.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190871.hg.1 ENST00000510971.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190871.hg.1 NR_039978.1 Refseq
          Affymetrix Human Clariom D Assay PSR0400190871.hg.1 TC0400012954.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0400190871.hg.1 ENST00000499359.2 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190871.hg.1 ENST00000510971.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190871.hg.1 ENST00000510971.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0400190871.hg.1 NR_039978.1 Refseq
          Human Genome U133 Plus 2.0 Array 1569713_PM_at - -
          Human Genome U133 Plus 2.0 Array 1569713_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569713_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569713_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569713_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569713_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569713_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569713_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569713_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569713_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569713_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569713_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1569713_at7 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A024604 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A024604 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix