Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04274777_g1
          See other RAD51-AS1 GE Assays ›
          Gene Symbol:
          RAD51-AS1
          Gene Name:
          RAD51 antisense RNA 1 (head to head)
          Gene Aliases:
          TODRA
          Chromosome Location:
          Chr.15: 40693734 - 40695105 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04274777_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RAD51-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040058.1 1 - 2 98 62
          GenBank mRNA AK125393.1 - 1 - 2 98 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1500009118.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC15001241.hg.1

          Set Membership:

          Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 581992 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 31909 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 148930 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P30831 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P30831 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 17433 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 48686 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P03014329 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P30831 - -
          SurePrint G3 Human GE Microarray G4851A A_32_P30831 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P30831 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03014329 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_18_P20288145 - -
          Agilent Human 1B Microarray G4111A A_32_P777021 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P03014329 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR15011190.hg.1 ENST00000526635.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15011190.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15011190.hg.1 NR_040058.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15011190.hg.1 TC15001241.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR15011191.hg.1 ENST00000526635.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15011191.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15011191.hg.1 NR_040058.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15011191.hg.1 TC15001241.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1500009118.hg.1 ENST00000499988.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009118.hg.1 ENST00000499988.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009118.hg.1 ENST00000499988.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009118.hg.1 ENST00000499988.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009118.hg.1 ENST00000499988.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009118.hg.1 ENST00000499988.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009118.hg.1 ENST00000499988.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009118.hg.1 ENST00000526635.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009118.hg.1 ENST00000526635.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009118.hg.1 ENST00000526635.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009118.hg.1 ENST00000526635.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009118.hg.1 ENST00000526635.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009118.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009118.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009118.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009118.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009118.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009118.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009118.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009118.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009118.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009118.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009118.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009118.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500009118.hg.1 NR_040058.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009118.hg.1 NR_040058.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009118.hg.1 NR_040058.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009118.hg.1 NR_040058.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009118.hg.1 NR_040058.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009118.hg.1 NR_040058.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500009118.hg.1 NR_040058.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15011189.hg.1 NR_040058.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15011189.hg.1 TC15001241.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1500165971.hg.1 ENST00000499988.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165971.hg.1 NR_040058.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500165971.hg.1 TC1500009118.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500165971.hg.1 NR_040058.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500165971.hg.1 ENST00000499988.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165972.hg.1 ENST00000499988.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165972.hg.1 NR_040058.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500165972.hg.1 TC1500009118.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500165972.hg.1 ENST00000499988.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165972.hg.1 NR_040058.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500165973.hg.1 ENST00000499988.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165973.hg.1 ENST00000526635.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165973.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165973.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165973.hg.1 NR_040058.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500165973.hg.1 TC1500009118.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500165973.hg.1 ENST00000499988.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165973.hg.1 ENST00000526635.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165973.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165973.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165973.hg.1 NR_040058.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500165976.hg.1 ENST00000499988.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165976.hg.1 ENST00000526635.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165976.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165976.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165976.hg.1 NR_040058.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500165976.hg.1 TC1500009118.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500165976.hg.1 ENST00000499988.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165976.hg.1 ENST00000526635.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165976.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165976.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165976.hg.1 NR_040058.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500165974.hg.1 ENST00000499988.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165974.hg.1 ENST00000526635.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165974.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165974.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165974.hg.1 NR_040058.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500165974.hg.1 TC1500009118.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500165974.hg.1 ENST00000499988.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165974.hg.1 ENST00000526635.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165974.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165974.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165974.hg.1 NR_040058.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500165975.hg.1 ENST00000499988.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165975.hg.1 ENST00000526635.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165975.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165975.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165975.hg.1 NR_040058.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500165975.hg.1 TC1500009118.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500165975.hg.1 ENST00000499988.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165975.hg.1 ENST00000526635.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165975.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165975.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165975.hg.1 NR_040058.1 Refseq
          Affymetrix Human Clariom D Assay TC1500009118.hg.1 ENST00000499988.2 Ensembl
          Affymetrix Human Clariom D Assay TC1500009118.hg.1 ENST00000499988.2 Ensembl
          Affymetrix Human Clariom D Assay TC1500009118.hg.1 ENST00000499988.2 Ensembl
          Affymetrix Human Clariom D Assay TC1500009118.hg.1 ENST00000499988.2 Ensembl
          Affymetrix Human Clariom D Assay TC1500009118.hg.1 ENST00000499988.2 Ensembl
          Affymetrix Human Clariom D Assay TC1500009118.hg.1 ENST00000499988.2 Ensembl
          Affymetrix Human Clariom D Assay TC1500009118.hg.1 ENST00000499988.2 Ensembl
          Affymetrix Human Clariom D Assay TC1500009118.hg.1 ENST00000526635.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009118.hg.1 ENST00000526635.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009118.hg.1 ENST00000526635.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009118.hg.1 ENST00000526635.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009118.hg.1 ENST00000526635.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009118.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009118.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009118.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009118.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009118.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009118.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009118.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009118.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009118.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009118.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009118.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009118.hg.1 ENST00000533146.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500009118.hg.1 NR_040058.1 Refseq
          Affymetrix Human Clariom D Assay TC1500009118.hg.1 NR_040058.1 Refseq
          Affymetrix Human Clariom D Assay TC1500009118.hg.1 NR_040058.1 Refseq
          Affymetrix Human Clariom D Assay TC1500009118.hg.1 NR_040058.1 Refseq
          Affymetrix Human Clariom D Assay TC1500009118.hg.1 NR_040058.1 Refseq
          Affymetrix Human Clariom D Assay TC1500009118.hg.1 NR_040058.1 Refseq
          Affymetrix Human Clariom D Assay TC1500009118.hg.1 NR_040058.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500165970.hg.1 ENST00000499988.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165970.hg.1 NR_040058.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500165970.hg.1 TC1500009118.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500165970.hg.1 ENST00000499988.2 Ensembl
          Affymetrix Human Clariom D Assay PSR1500165970.hg.1 NR_040058.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A052022 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A052022 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0