Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04274807_m1
          See other LOC643339 GE Assays ›
          Gene Symbol:
          LOC643339
          Gene Name:
          uncharacterized LOC643339
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 93003758 - 93377736 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04274807_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC643339
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040096.1 4 - 5 509 124
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CD684822.1 - 2 - 3 190 Chr. 12 - 93003496 - 93139132 [-] on Build GRCh38 124
          GenBankmRNA BU659324.1 - 4 - 5 509 Chr. 12 - 93003758 - 93377730 [-] on Build GRCh38 124

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream early endosome antigen 1 EEA1 MST105,
          MSTP105,
          ZFYVE2
          -74.4kb Sense Chr. 12 - 92772509 - 92975228 on NCBI Build GRCh38
          Downstream ribosomal protein L41 pseudogene 5 RPL41P5 RPL41L 78.7kb Sense Chr. 12 - 93083293 - 93083716 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1200011526.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC12001812.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3332533 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR12023944.hg.1 ENST00000549930.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12023944.hg.1 ENST00000550324.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12023944.hg.1 NR_040096.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12023944.hg.1 TC12001812.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR12023945.hg.1 ENST00000549930.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12023945.hg.1 ENST00000550324.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12023945.hg.1 NR_040096.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12023945.hg.1 TC12001812.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR12023953.hg.1 ENST00000547432.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12023953.hg.1 ENST00000549930.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12023953.hg.1 NR_040096.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12023953.hg.1 TC12001812.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR12023957.hg.1 ENST00000548890.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12023957.hg.1 ENST00000549806.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12023957.hg.1 ENST00000552835.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12023957.hg.1 NR_040096.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12023957.hg.1 TC12001812.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR12023959.hg.1 ENST00000548890.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12023959.hg.1 ENST00000549806.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12023959.hg.1 ENST00000552835.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR12023959.hg.1 NR_040096.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR12023959.hg.1 TC12001812.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000547432.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000547432.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000547432.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000547432.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000547432.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000548890.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000548890.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000549806.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000549806.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000549806.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000549930.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000549930.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000549930.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000550324.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000550324.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000550324.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000550324.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000550324.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000550324.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000550324.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000550324.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000552100.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000552100.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000552451.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000552451.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000552451.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000552835.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000552835.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000552835.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000552835.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000552835.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000552835.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000552835.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000552835.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000552835.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 ENST00000552835.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 NR_040096.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 NR_040096.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 NR_040096.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 NR_040096.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1200011526.hg.1 NR_040096.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200189334.hg.1 ENST00000549930.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200189334.hg.1 ENST00000550324.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200189334.hg.1 ENST00000550324.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200189334.hg.1 NR_040096.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200189334.hg.1 TC1200011526.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200189334.hg.1 NR_040096.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200189334.hg.1 ENST00000549930.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200189334.hg.1 ENST00000550324.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200189334.hg.1 ENST00000550324.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200189353.hg.1 ENST00000547432.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200189353.hg.1 ENST00000549930.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200189353.hg.1 NR_040096.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200189353.hg.1 TC1200011526.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200189353.hg.1 ENST00000547432.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200189353.hg.1 ENST00000549930.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200189353.hg.1 NR_040096.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200189358.hg.1 ENST00000548890.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200189358.hg.1 ENST00000549806.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200189358.hg.1 ENST00000552835.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200189358.hg.1 ENST00000552835.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200189358.hg.1 NR_040096.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200189358.hg.1 TC1200011526.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200189358.hg.1 ENST00000548890.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200189358.hg.1 ENST00000549806.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200189358.hg.1 ENST00000552835.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200189358.hg.1 ENST00000552835.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200189358.hg.1 NR_040096.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200189335.hg.1 ENST00000549930.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200189335.hg.1 ENST00000550324.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200189335.hg.1 ENST00000550324.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200189335.hg.1 NR_040096.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200189335.hg.1 TC1200011526.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200189335.hg.1 ENST00000549930.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200189335.hg.1 ENST00000550324.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200189335.hg.1 ENST00000550324.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200189335.hg.1 NR_040096.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200189360.hg.1 ENST00000548890.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200189360.hg.1 ENST00000549806.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200189360.hg.1 ENST00000552835.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200189360.hg.1 ENST00000552835.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200189360.hg.1 NR_040096.1 Refseq
          Affymetrix Human Clariom D Assay PSR1200189360.hg.1 TC1200011526.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1200189360.hg.1 ENST00000548890.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200189360.hg.1 ENST00000549806.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200189360.hg.1 ENST00000552835.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200189360.hg.1 ENST00000552835.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1200189360.hg.1 NR_040096.1 Refseq
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000547432.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000547432.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000547432.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000547432.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000547432.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000548890.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000548890.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000549806.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000549806.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000549806.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000549930.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000549930.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000549930.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000550324.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000550324.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000550324.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000550324.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000550324.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000550324.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000550324.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000550324.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000552100.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000552100.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000552451.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000552451.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000552451.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000552835.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000552835.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000552835.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000552835.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000552835.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000552835.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000552835.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000552835.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000552835.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 ENST00000552835.1 Ensembl
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 NR_040096.1 Refseq
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 NR_040096.1 Refseq
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 NR_040096.1 Refseq
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 NR_040096.1 Refseq
          Affymetrix Human Clariom D Assay TC1200011526.hg.1 NR_040096.1 Refseq
          Human Genome U95 Set 68377_PM_at - -
          Human Genome U95 Set 68377_at - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1727852 NR_040096.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1727852 NR_040096.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1727852 NR_040096.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1727852 NR_040096.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1727852 NR_040096.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0