Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04274813_m1
          See other ADPGK-AS1 GE Assays ›
          Gene Symbol:
          ADPGK-AS1
          Gene Name:
          ADPGK antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 72782835 - 72798199 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04274813_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ADPGK-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040107.1 1 - 2 2359 89
          GenBank mRNA AK127877.1 - 1 - 2 2361 89
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DB330252.1 - 2 - 3 Chr. 15 - 72785031 - 72798199 [-] on Build GRCh38 89
          GenBankmRNA CA306872.1 - 2 - 3 Chr. 15 - 72785012 - 72792963 [-] on Build GRCh38 89

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream ADP dependent glucokinase ADPGK 2610017G09Rik,
          ADP-GK
          Anti-sense Chr. 15 - 72751367 - 72783785 on NCBI Build GRCh38
          Downstream neogenin 1 NEO1 IGDCC2,
          NGN,
          NTN1R2
          264.1kb Sense Chr. 15 - 73051715 - 73305206 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1500007806.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC15000663.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30246595 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09752888 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30246626 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09752908 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30246599 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30246592 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30246605 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09752890 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30246630 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30246622 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30246600 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09752902 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09752899 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09752919 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09752918 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30246623 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09752929 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09752898 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09752905 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09752923 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30246625 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09752927 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09752931 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09752891 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09752915 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30246635 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30246593 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09752909 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09752904 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09752911 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30246591 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30246610 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09752930 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30246604 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09752912 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30246615 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09752903 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09752914 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30246598 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30246594 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P30246621 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P09752922 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 802765 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 772935 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 140517 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 773565 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 636811 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 139409 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P57328785 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03060001 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P20301286 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P20301296 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P20301290 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03059996 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03060001 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P03060001 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR15005698.hg.1 NR_040107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15005698.hg.1 TC15000663.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR15005699.hg.1 NR_040107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15005699.hg.1 TC15000663.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR15005700.hg.1 NR_040107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15005700.hg.1 TC15000663.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC1500007806.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007806.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007806.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007806.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007806.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007806.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007806.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007806.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007806.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007806.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007806.hg.1 ENST00000566745.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007806.hg.1 ENST00000566745.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007806.hg.1 ENST00000566745.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007806.hg.1 ENST00000566745.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1500007806.hg.1 NR_040107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007806.hg.1 NR_040107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007806.hg.1 NR_040107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007806.hg.1 NR_040107.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1500007806.hg.1 NR_040107.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500155424.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500155424.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500155424.hg.1 NR_040107.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500155424.hg.1 TC1500007806.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500155424.hg.1 NR_040107.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500155424.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500155424.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007806.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007806.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007806.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007806.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007806.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007806.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007806.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007806.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007806.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007806.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007806.hg.1 ENST00000566745.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007806.hg.1 ENST00000566745.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007806.hg.1 ENST00000566745.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007806.hg.1 ENST00000566745.1 Ensembl
          Affymetrix Human Clariom D Assay TC1500007806.hg.1 NR_040107.1 Refseq
          Affymetrix Human Clariom D Assay TC1500007806.hg.1 NR_040107.1 Refseq
          Affymetrix Human Clariom D Assay TC1500007806.hg.1 NR_040107.1 Refseq
          Affymetrix Human Clariom D Assay TC1500007806.hg.1 NR_040107.1 Refseq
          Affymetrix Human Clariom D Assay TC1500007806.hg.1 NR_040107.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500155428.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500155428.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500155428.hg.1 ENST00000566745.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500155428.hg.1 NR_040107.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500155428.hg.1 TC1500007806.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500155428.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500155428.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500155428.hg.1 ENST00000566745.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500155428.hg.1 NR_040107.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500155423.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500155423.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500155423.hg.1 NR_040107.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500155423.hg.1 TC1500007806.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500155423.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500155423.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500155423.hg.1 NR_040107.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500155425.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500155425.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500155425.hg.1 ENST00000566745.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500155425.hg.1 NR_040107.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500155425.hg.1 TC1500007806.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500155425.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500155425.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500155425.hg.1 ENST00000566745.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500155425.hg.1 NR_040107.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500155432.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500155432.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500155432.hg.1 NR_040107.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500155432.hg.1 TC1500007806.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500155432.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500155432.hg.1 ENST00000563592.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500155432.hg.1 NR_040107.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A053047 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A053047 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-ztd5s:80/100.66.134.6:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0