Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04275781_s1
          See other PTOV1-AS1 GE Assays ›
          Gene Symbol:
          PTOV1-AS1 MED25
          Gene Name:
          PTOV1 antisense RNA 1
          mediator complex subunit 25
          Gene Aliases:
          -
          ACID1, ARC92, BVSYS, CMT2B2, P78, PTOV2, TCBAP0758
          Chromosome Location:
          Chr.19: 49838645 - 49851676 on Build GRCh38
          Chromosome Location:
          Chr.19: 49818279 - 49840384 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs04275781_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PTOV1-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040037.1 4 2021 113
          GenBank mRNA BC043359.1 - 4 2021 113
          Gene Symbol: MED25
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_011527353.1 XP_011525655.1 18 113
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CN284979.1 - 1 Chr. 19 - 49838537 - 49839326 [+] on Build GRCh38 113
          GenBankmRNA BU169402.1 - 1 Chr. 19 - 49838602 - 49839334 [+] on Build GRCh38 113
          GenBankmRNA BE154251.1 - 1 Chr. 19 - 49838533 - 49839095 [+] on Build GRCh38 113
          GenBankmRNA BM564251.1 - 3 Chr. 19 - 49835845 - 49839354 [+] on Build GRCh38 113
          GenBankmRNA AW872394.1 - 1 Chr. 19 - 49838647 - 49839145 [+] on Build GRCh38 113

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream microRNA 6800 MIR6800 hsa-mir-6800 -6.5kb Anti-sense Chr. 19 - 49832018 - 49832099 on NCBI Build GRCh38
          Downstream prostate tumor overexpressed 1 PTOV1 ACID2,
          PTOV-1
          8.8kb Anti-sense Chr. 19 - 49850726 - 49860744 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1900011165.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC19001726.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          transcription initiation from RNA polymerase II promoter
          positive regulation of chromatin binding
          positive regulation of transcription by RNA polymerase II
          negative regulation of fibroblast proliferation
          positive regulation of mediator complex assembly
          protein binding
          transcription factor binding
          retinoic acid receptor binding
          retinoid X receptor binding

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P21028654 - -
          Agilent Human 1B Microarray G4111A A_32_P753489 - -
          Agilent Human 1B Microarray G4111A A_32_P7426 - -
          Agilent Human 1B Microarray G4111A A_32_P378673 - -
          Agilent Human 1B Microarray G4111A A_32_P803469 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR19023566.hg.1 TC19001726.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19023566.hg.1 NR_040037.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19023564.hg.1 TC19001726.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19023564.hg.1 NR_040037.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19023563.hg.1 TC19001726.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19023563.hg.1 NR_040037.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR19023565.hg.1 TC19001726.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR19023565.hg.1 NR_040037.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011165.hg.1 ENST00000596521.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011165.hg.1 ENST00000596521.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011165.hg.1 ENST00000596521.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011165.hg.1 ENST00000596521.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011165.hg.1 ENST00000596521.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011165.hg.1 ENST00000596521.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011165.hg.1 ENST00000596521.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011165.hg.1 ENST00000600742.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011165.hg.1 ENST00000600742.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011165.hg.1 ENST00000600742.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011165.hg.1 ENST00000600742.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900011165.hg.1 NR_040037.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011165.hg.1 NR_040037.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011165.hg.1 NR_040037.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011165.hg.1 NR_040037.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011165.hg.1 NR_040037.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011165.hg.1 NR_040037.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900011165.hg.1 NR_040037.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900193595.hg.1 ENST00000596521.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900193595.hg.1 ENST00000600742.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900193595.hg.1 NR_040037.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900193595.hg.1 TC1900011165.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900193595.hg.1 NR_040037.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900193595.hg.1 ENST00000596521.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900193595.hg.1 ENST00000600742.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900193596.hg.1 ENST00000596521.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900193596.hg.1 NR_040037.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900193596.hg.1 TC1900011165.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900193596.hg.1 ENST00000596521.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900193596.hg.1 NR_040037.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900193592.hg.1 ENST00000596521.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900193592.hg.1 NR_040037.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900193592.hg.1 TC1900011165.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900193592.hg.1 ENST00000596521.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900193592.hg.1 NR_040037.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900193599.hg.1 ENST00000596521.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900193599.hg.1 NR_040037.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900193599.hg.1 TC1900011165.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900193599.hg.1 ENST00000596521.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900193599.hg.1 NR_040037.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011165.hg.1 ENST00000596521.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011165.hg.1 ENST00000596521.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011165.hg.1 ENST00000596521.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011165.hg.1 ENST00000596521.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011165.hg.1 ENST00000596521.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011165.hg.1 ENST00000596521.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011165.hg.1 ENST00000596521.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011165.hg.1 ENST00000600742.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011165.hg.1 ENST00000600742.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011165.hg.1 ENST00000600742.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011165.hg.1 ENST00000600742.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900011165.hg.1 NR_040037.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011165.hg.1 NR_040037.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011165.hg.1 NR_040037.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011165.hg.1 NR_040037.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011165.hg.1 NR_040037.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011165.hg.1 NR_040037.1 Refseq
          Affymetrix Human Clariom D Assay TC1900011165.hg.1 NR_040037.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900193593.hg.1 ENST00000596521.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900193593.hg.1 ENST00000600742.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900193593.hg.1 NR_040037.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900193593.hg.1 TC1900011165.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900193593.hg.1 ENST00000596521.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900193593.hg.1 ENST00000600742.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900193593.hg.1 NR_040037.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900193597.hg.1 ENST00000596521.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900193597.hg.1 NR_040037.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900193597.hg.1 TC1900011165.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900193597.hg.1 ENST00000596521.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900193597.hg.1 NR_040037.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900193603.hg.1 ENST00000596521.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900193603.hg.1 NR_040037.1 Refseq
          Affymetrix Human Clariom D Assay PSR1900193603.hg.1 TC1900011165.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1900193603.hg.1 ENST00000596521.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1900193603.hg.1 NR_040037.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A011699 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A011699 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1830842 AK092213.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1830842 BC043359.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1830842 NR_040037.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1830842 AK092213.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1830842 BC043359.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_1830842 NR_040037.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1830842 AK092213.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1830842 BC043359.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1830842 NR_040037.1 human_transcriptome
          Illumina Sentrix Human-6 v2 Expression BeadChip 5260050 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_117725 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1830842 AK092213.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1830842 BC043359.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1830842 NR_040037.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1830842 AK092213.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1830842 BC043359.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1830842 NR_040037.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1830842 AK092213.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1830842 BC043359.1 human_transcriptome
          Illumina HumanWG-6_V2_0_R2 ILMN_1830842 NR_040037.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0