Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04331879_m1
          See other CECR3 GE Assays ›
          Gene Symbol:
          CECR3
          Gene Name:
          cat eye syndrome chromosome region, candidate 3 (non-protein coding)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.22: 17256859 - 17266733 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04331879_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CECR3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038398.2 2 - 3 469 82
          GenBank mRNA AF277398.1 - 2 - 3 469 82

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream cat eye syndrome chromosome region,
          candidate 1
          CECR1 ADA2,
          ADGF,
          IDGFL,
          PAN,
          SNEDS
          -35.0kb Sense Chr. 22 - 17178790 - 17221854 on NCBI Build GRCh38
          Downstream CECR2,
          histone acetyl-lysine reader
          CECR2 98.9kb Anti-sense Chr. 22 - 17359949 - 17558155 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2200007896.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC22001479.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423468 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32091726 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423489 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423490 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32091714 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423514 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423531 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423515 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17251426 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32091739 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17251427 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423526 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423507 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423546 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423528 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17251457 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32091773 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17251431 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423480 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423501 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423551 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32091697 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423519 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423484 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17251456 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32091797 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423535 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423474 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423472 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32091727 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423494 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423532 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32091685 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32091700 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423538 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423457 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17251435 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17251459 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32091749 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423481 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423478 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32091791 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423555 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423498 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423525 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32091767 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32091695 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32091770 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423479 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17251446 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32091690 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32091799 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423486 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423500 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423473 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423467 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423455 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423541 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423557 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32091792 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32091758 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17251443 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32091757 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423509 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32091730 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423511 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32091740 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423510 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32091759 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423459 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32091793 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423463 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32091746 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17251437 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17251432 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423537 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423536 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423470 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423476 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32091718 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423452 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423524 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423508 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423542 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423548 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32091717 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423461 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17251449 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423497 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423520 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17251447 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17251436 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423502 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423450 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423471 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423492 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32091803 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32091735 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423482 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423462 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17251463 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32091708 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P32091788 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423518 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423505 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11423554 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 133579 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 565433 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 480522 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 413487 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 487584 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 261680 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 93250 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3481113 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 15061 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3481113 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3481113 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P03598970 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P21275890 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P03598970 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P21275890 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P03598970 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 1564352_PM_at - -
          Affymetrix Human Transcriptome Array 2.0 PSR22008476.hg.1 TC22001479.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22008476.hg.1 ENST00000428828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22008476.hg.1 NR_038398.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22008480.hg.1 TC22001479.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22008480.hg.1 ENST00000428828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22008480.hg.1 NR_038398.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22008478.hg.1 TC22001479.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22008478.hg.1 ENST00000428828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22008478.hg.1 NR_038398.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22008479.hg.1 TC22001479.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22008479.hg.1 ENST00000428828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22008479.hg.1 NR_038398.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR22008481.hg.1 TC22001479.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR22008481.hg.1 ENST00000428828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR22008481.hg.1 NR_038398.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007896.hg.1 ENST00000428828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007896.hg.1 ENST00000428828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007896.hg.1 ENST00000428828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007896.hg.1 ENST00000428828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007896.hg.1 ENST00000428828.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2200007896.hg.1 NR_038398.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007896.hg.1 NR_038398.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007896.hg.1 NR_038398.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007896.hg.1 NR_038398.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2200007896.hg.1 NR_038398.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007896.hg.1 ENST00000428828.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200007896.hg.1 ENST00000428828.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200007896.hg.1 ENST00000428828.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200007896.hg.1 ENST00000428828.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200007896.hg.1 ENST00000428828.1 Ensembl
          Affymetrix Human Clariom D Assay TC2200007896.hg.1 NR_038398.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007896.hg.1 NR_038398.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007896.hg.1 NR_038398.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007896.hg.1 NR_038398.2 Refseq
          Affymetrix Human Clariom D Assay TC2200007896.hg.1 NR_038398.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200156132.hg.1 ENST00000428828.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200156132.hg.1 NR_038398.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200156132.hg.1 TC2200007896.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200156132.hg.1 ENST00000428828.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200156132.hg.1 NR_038398.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200156134.hg.1 ENST00000428828.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200156134.hg.1 NR_038398.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200156134.hg.1 TC2200007896.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200156134.hg.1 ENST00000428828.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200156134.hg.1 NR_038398.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200156130.hg.1 ENST00000428828.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200156130.hg.1 NR_038398.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200156130.hg.1 TC2200007896.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200156130.hg.1 ENST00000428828.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200156130.hg.1 NR_038398.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200156131.hg.1 ENST00000428828.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200156131.hg.1 NR_038398.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200156131.hg.1 TC2200007896.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200156131.hg.1 ENST00000428828.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200156131.hg.1 NR_038398.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200156133.hg.1 ENST00000428828.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200156133.hg.1 NR_038398.2 Refseq
          Affymetrix Human Clariom D Assay PSR2200156133.hg.1 TC2200007896.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2200156133.hg.1 ENST00000428828.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2200156133.hg.1 NR_038398.2 Refseq
          Human Genome U133 Plus 2.0 Array 1564352_at - -
          Human Genome U133 Plus 2.0 Array 1564352_PM_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564352_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564352_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564352_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564352_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564352_at9 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564352_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564352_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564352_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564352_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564352_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 1564352_at11 - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A019435 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A019435 - -
          Array Name Probe IDs Targets Target Type
          Illumina human-6 v2.0 expression beadchip ILMN_1854474 - -
          Illumina Sentrix Human-6 v2 Expression BeadChip 6840398 - -
          Illumina HumanRef-6 v2.0 expression beadchip ILMN_99246 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1854474 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1854474 - -
          Illumina HumanWG-6_V2_0_R2 ILMN_1854474 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0