Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04331888_m1
          See other LOC643441 GE Assays ›
          Gene Symbol:
          LOC643441
          Gene Name:
          uncharacterized LOC643441
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 113196781 - 113206253 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04331888_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC643441
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_038846.1 2 - 3 261 120
          GenBank mRNA AK123703.1 - 2 - 3 261 120
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DA589239.1 - 2 - 3 261 Chr. 1 - 113198396 - 113206253 [-] on Build GRCh38 120

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC102723919 LOC102723919 -64.5kb Anti-sense Chr. 1 - 113131186 - 113132260 on NCBI Build GRCh38
          Downstream membrane associated guanylate kinase,
          WW and PDZ domain containing 3
          MAGI3 MAGI-3,
          dJ730K3.2
          188.6kb Anti-sense Chr. 1 - 113390474 - 113685923 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100015321.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01003003.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Homo sapiens 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465826 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465782 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20498639 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20498713 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465822 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465873 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465867 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465833 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465816 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465810 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20498650 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465835 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465874 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20498736 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465805 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465868 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P15101002 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20498657 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465860 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465866 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20498689 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465875 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465825 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20498668 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465802 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20498666 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465840 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465784 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20498703 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20498696 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465871 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465801 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20498646 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465862 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465855 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20498681 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465881 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465814 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465883 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465811 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20498640 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20498723 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465786 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20498716 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20498699 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465870 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465789 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465788 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465804 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465857 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465792 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20498656 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20498693 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465783 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465809 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465876 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465830 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20498726 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465820 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20498747 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465799 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465845 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465808 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20498707 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20498637 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465887 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20498701 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465797 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465846 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20498698 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465798 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20498711 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20498714 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465882 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20498641 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465847 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465842 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465872 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465856 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465843 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465878 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20498688 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20498715 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20498721 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465823 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465841 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20498655 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P20498649 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P00465821 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 122190 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 94263 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 806212 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3401307 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P9151 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 297 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 16989 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3401307 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3401307 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P9151 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 151843 AB019524.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 151843 AB028970.2 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 151843 AF044209.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 151843 AF087856.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 151843 AF303584.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 151843 AF303585.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 151843 AK310405.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 151843 AL137641.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 151843 CR625140.1 genbank_mrna_human
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 151843 NM_006311.2 refseq_mrna_human_NM
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15263017 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P15263018 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P00149980 - -
          Agilent Human 1B Microarray G4111A A_32_P9151 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P00149983 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR01046676.hg.1 NR_038846.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01046676.hg.1 TC01003003.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR0100214292.hg.1 NR_038846.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100214292.hg.1 TC0100015321.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100214292.hg.1 NR_038846.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100214293.hg.1 NR_038846.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100214293.hg.1 TC0100015321.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100214293.hg.1 NR_038846.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100214294.hg.1 NR_038846.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100214294.hg.1 TC0100015321.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100214294.hg.1 NR_038846.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100214295.hg.1 NR_038846.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100214295.hg.1 TC0100015321.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100214295.hg.1 NR_038846.1 Refseq
          Human Genome U95 Set 87437_PM_at - -
          Human Genome U95 Set 87437_at - -
          Human Genome U95 Set 76373_PM_at - -
          Human Genome U95 Set 76373_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A049804 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A049804 - -
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 144095 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1751558 - -
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1751558 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1751558 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1751558 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1751558 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0