Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04332293_s1
          See other NIPBL-AS1 GE Assays ›
          Gene Symbol:
          NIPBL-AS1
          Gene Name:
          NIPBL antisense RNA 1 (head to head)
          Gene Aliases:
          NIPBL-AS
          Chromosome Location:
          Chr.5: 36871361 - 36876694 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04332293_s1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NIPBL-AS1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_046262.1 1 5219 80
          GenBank mRNA AI800212.1 - 1 80
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CX782531.1 - 1 Chr. 5 - 36871364 - 36872066 [+] on Build GRCh38 80
          GenBankmRNA BE350752.1 - 1 Chr. 5 - 36871361 - 36871823 [+] on Build GRCh38 80
          GenBankmRNA AV753237.1 - 1 781 Chr. 5 - 36871361 - 36872106 [-] on Build GRCh38 80
          GenBankmRNA BF734993.1 - 1 Chr. 5 - 36871151 - 36871559 [+] on Build GRCh38 80
          GenBankmRNA BX106017.1 - 1 Chr. 5 - 36871361 - 36871855 [+] on Build GRCh38 80
          GenBankmRNA AI873320.1 - 1 Chr. 5 - 36871361 - 36871620 [+] on Build GRCh38 80
          GenBankmRNA AI286236.1 - 1 Chr. 5 - 36871361 - 36871798 [+] on Build GRCh38 80
          GenBankmRNA DB309142.1 - 1 Chr. 5 - 36871364 - 36871911 [+] on Build GRCh38 80
          GenBankmRNA AI921947.1 - 1 Chr. 5 - 36871361 - 36871799 [+] on Build GRCh38 80
          GenBankmRNA AI051298.1 - 1 Chr. 5 - 36871361 - 36871729 [+] on Build GRCh38 80
          GenBankmRNA DB528963.1 - 1 354 Chr. 5 - 36871369 - 36871830 [-] on Build GRCh38 80

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream solute carrier family 1 member 3 SLC1A3 EA6,
          EAAT1,
          GLAST,
          GLAST1
          -183.0kb Anti-sense Chr. 5 - 36606355 - 36688334 on NCBI Build GRCh38
          Downstream NIPBL,
          cohesin loading factor
          NIPBL CDLS,
          CDLS1,
          IDN3,
          IDN3-B,
          Scc2
          65.0 Anti-sense Chr. 5 - 36876759 - 37065819 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0500010508.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24047047 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24047066 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24047039 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03875198 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24047035 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03875230 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03875236 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03875213 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03875197 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24047056 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24047069 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24047049 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03875223 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24047042 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03875234 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03875243 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24047054 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24047030 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24047052 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03875216 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24047029 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24047061 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24047045 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24047022 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24047062 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24047043 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03875225 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24047034 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03875205 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24047033 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03875241 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03875194 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03875219 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03875199 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24047041 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24047067 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24047028 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P03875244 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P24047050 - -
          Agilent SurePrint G3 Human CGH Microarray 4x180K 143566 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 933038 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 609824 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 522157 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 240712 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 590855 - -
          Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k GT_44k_32_P126410 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 9099 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 46626 - -
          Agilent Human Genome CGH + SNP Microarray A_16_P17089989 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3419865 - -
          Whole Human Genome Oligo Microarray G4112A A_32_P126410 - -
          Agilent Human Genome 244K CGH Microarray (Alpha Test) A_16_P17089989 - -
          Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 104195 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P37149761 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P17089976 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01213878 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P17089989 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P01213873 - -
          Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) A_16_P37149727 - -
          Agilent Human 1B Microarray G4111A A_32_P126410 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P17089989 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P37149727 - -
          Agilent SurePrint G3 Human CGH Microarray 2x400K A_16_P01213878 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Clariom D Assay PSR0500173174.hg.1 NR_046262.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500173174.hg.1 TC0500010508.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500173174.hg.1 NR_046262.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500173172.hg.1 NR_046262.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500173172.hg.1 TC0500010508.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500173172.hg.1 NR_046262.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500173173.hg.1 NR_046262.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500173173.hg.1 TC0500010508.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500173173.hg.1 NR_046262.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500173175.hg.1 NR_046262.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500173175.hg.1 TC0500010508.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500173175.hg.1 NR_046262.1 Refseq
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1751847 - -
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1751847 - -
          Illumina human-6 v2.0 expression beadchip ILMN_1751847 - -
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1751847 - -
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1751847 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0