Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010251.hg.1 |
ENST00000414969.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010251.hg.1 |
ENST00000414969.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010251.hg.1 |
ENST00000414969.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010251.hg.1 |
NR_046647.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010251.hg.1 |
NR_046647.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300010251.hg.1 |
NR_046647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010251.hg.1 |
ENST00000414969.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010251.hg.1 |
ENST00000414969.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010251.hg.1 |
ENST00000414969.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300010251.hg.1 |
NR_046647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010251.hg.1 |
NR_046647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300010251.hg.1 |
NR_046647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300174862.hg.1 |
ENST00000414969.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300174862.hg.1 |
NR_046647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300174862.hg.1 |
TC0300010251.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300174862.hg.1 |
NR_046647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300174862.hg.1 |
ENST00000414969.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300174861.hg.1 |
ENST00000414969.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300174861.hg.1 |
NR_046647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300174861.hg.1 |
TC0300010251.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300174861.hg.1 |
ENST00000414969.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300174861.hg.1 |
NR_046647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300174860.hg.1 |
ENST00000414969.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300174860.hg.1 |
NR_046647.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300174860.hg.1 |
TC0300010251.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300174860.hg.1 |
ENST00000414969.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300174860.hg.1 |
NR_046647.1 |
Refseq |