Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010173.hg.1 |
ENST00000443837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010173.hg.1 |
ENST00000443837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010173.hg.1 |
ENST00000443837.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010173.hg.1 |
NR_046834.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010173.hg.1 |
NR_046834.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700010173.hg.1 |
NR_046834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010173.hg.1 |
ENST00000443837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010173.hg.1 |
ENST00000443837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010173.hg.1 |
ENST00000443837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700010173.hg.1 |
NR_046834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010173.hg.1 |
NR_046834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700010173.hg.1 |
NR_046834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700172314.hg.1 |
ENST00000443837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700172314.hg.1 |
NR_046834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700172314.hg.1 |
TC0700010173.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700172314.hg.1 |
NR_046834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700172314.hg.1 |
ENST00000443837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700172313.hg.1 |
ENST00000443837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700172313.hg.1 |
NR_046834.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700172313.hg.1 |
TC0700010173.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700172313.hg.1 |
ENST00000443837.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700172313.hg.1 |
NR_046834.1 |
Refseq |