Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04332609_g1
          See other SNX17 GE Assays ›
          Gene Symbol:
          SNX17
          Gene Name:
          sorting nexin 17
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 27370496 - 27377533 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04332609_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SNX17
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_049783.1 3 - 4 524 70
          NR_049784.1 3 - 4 500 70
          GenBank mRNA AK316373.1 - 2 - 3 205 70
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA DA263252.1 - 3 - 4 362 Chr. 2 - 27370636 - 27373924 [+] on Build GRCh38 70
          GenBankmRNA DC395134.1 - 2 - 3 205 Chr. 2 - 27371253 - 27374171 [+] on Build GRCh38 70
          GenBankmRNA BQ224277.1 - 3 - 4 348 Chr. 2 - 27370648 - 27374744 [+] on Build GRCh38 70
          GenBankmRNA BP379371.1 - 3 - 4 378 Chr. 2 - 27370617 - 27373892 [+] on Build GRCh38 70
          GenBankmRNA DA259406.1 - 3 - 4 379 Chr. 2 - 27370617 - 27373883 [+] on Build GRCh38 70
          GenBankmRNA DB029853.1 - 3 - 4 417 Chr. 2 - 27370579 - 27373896 [+] on Build GRCh38 70
          GenBankmRNA BP341199.1 - 3 - 4 375 Chr. 2 - 27370620 - 27373924 [+] on Build GRCh38 70
          GenBankmRNA DA876290.1 - 3 - 4 376 Chr. 2 - 27370620 - 27373904 [+] on Build GRCh38 70
          GenBankmRNA BG437427.1 - 3 - 4 369 Chr. 2 - 27370628 - 27374393 [+] on Build GRCh38 70
          GenBankmRNA DA946195.1 - 3 - 4 376 Chr. 2 - 27370620 - 27373893 [+] on Build GRCh38 70
          GenBankmRNA DA528095.1 - 3 - 4 376 Chr. 2 - 27370620 - 27373944 [+] on Build GRCh38 70
          GenBankmRNA DB032395.1 - 3 - 4 379 Chr. 2 - 27370617 - 27373927 [+] on Build GRCh38 70

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream eukaryotic translation initiation factor 2B subunit delta EIF2B4 EIF-2B,
          EIF2B,
          EIF2Bdelta
          -2.1kb Anti-sense Chr. 2 - 27364352 - 27370457 on NCBI Build GRCh38
          Downstream zinc finger protein 513 ZNF513 HMFT0656,
          RP58
          4.1kb Anti-sense Chr. 2 - 27377231 - 27380866 on NCBI Build GRCh38

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK316373.1 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          cardiac septum development
          cholesterol catabolic process
          intracellular protein transport
          receptor-mediated endocytosis
          signal transduction
          endosomal transport
          regulation of endocytosis
          endocytic recycling
          aorta development
          coronary vasculature development
          signaling receptor binding
          protein binding
          protein C-terminus binding
          phosphatidylinositol binding
          low-density lipoprotein particle receptor binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0