Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04332669_g1
          See other LINC00462 GE Assays ›
          Gene Symbol:
          LINC00462
          Gene Name:
          long intergenic non-protein coding RNA 462
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 48576974 - 48580901 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04332669_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LINC00462
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_051983.1 2 - 3 132 93
          GenBank mRNA BM472085.1 - 2 - 3 133 93

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RCC1 and BTB domain containing protein 2 RCBTB2 CHC1L,
          RLG
          -43.7kb Sense Chr. 13 - 48488963 - 48535995 on NCBI Build GRCh38
          Downstream cysteinyl leukotriene receptor 2 CYSLTR2 CYSLT2,
          CYSLT2R,
          GPCR21,
          HG57,
          HPN321,
          KPG_011,
          PSEC0146,
          hGPCR21
          128.7kb Anti-sense Chr. 13 - 48653711 - 48709362 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1300008899.hg.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3326767 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1300008899.hg.1 ENST00000425350.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008899.hg.1 ENST00000425350.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1300008899.hg.1 NR_051983.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008899.hg.1 NR_051983.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1300008899.hg.1 NR_051983.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300160088.hg.1 ENST00000425350.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300160088.hg.1 NR_051983.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300160088.hg.1 TC1300008899.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300160088.hg.1 ENST00000425350.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300160088.hg.1 NR_051983.1 Refseq
          Affymetrix Human Clariom D Assay TC1300008899.hg.1 ENST00000425350.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300008899.hg.1 ENST00000425350.1 Ensembl
          Affymetrix Human Clariom D Assay TC1300008899.hg.1 NR_051983.1 Refseq
          Affymetrix Human Clariom D Assay TC1300008899.hg.1 NR_051983.1 Refseq
          Affymetrix Human Clariom D Assay TC1300008899.hg.1 NR_051983.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300160089.hg.1 ENST00000425350.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300160089.hg.1 NR_051983.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300160089.hg.1 TC1300008899.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300160089.hg.1 ENST00000425350.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1300160089.hg.1 NR_051983.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300160090.hg.1 NR_051983.1 Refseq
          Affymetrix Human Clariom D Assay PSR1300160090.hg.1 TC1300008899.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1300160090.hg.1 NR_051983.1 Refseq
          Human Genome U133 Plus 2.0 Array 240058_PM_at - -
          Human Genome U133 Plus 2.0 Array 240058_at - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240058_at10 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240058_at5 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240058_at4 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240058_at11 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240058_at2 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240058_at3 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240058_at7 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240058_at8 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240058_at1 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240058_at6 - -
          Affymetrix Human Genome U133 Plus 2.0 Array 240058_at9 - -
          Human Genome U95 Set 80782_at - -
          Human Genome U95 Set 80782_PM_at - -
          Human Genome U133 Set 240058_at - -
          Human Genome U133 Set 240058_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0