Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04333048_m1
          See other ADARB1 GE Assays ›
          Gene Symbol:
          ADARB1
          Gene Name:
          adenosine deaminase, RNA specific B1
          Gene Aliases:
          ADAR2, DRABA2, DRADA2, RED1
          Chromosome Location:
          Chr.21: 45074578 - 45226563 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04333048_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: ADARB1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027672.1 10 - 11 2982 78
          NR_027673.1 12 - 13 4035 78
          NR_027674.1 11 - 12 3917 78
          NR_073200.1 12 - 13 4037 78
          GenBank mRNA AB194369.1 - 10 - 11 2961 78
          AB194371.1 - 11 - 12 3896 78
          AF525422.1 - 12 - 13 4013 78
          AF533142.1 - 11 - 12 3895 78
          AY135659.1 - 12 - 13 4013 78
          U76421.1 - 12 - 13 4016 78
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BQ955425.1 - 1 - 2 128 Chr. 21 - 45223446 - 45226262 [+] on Build GRCh38 78

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC105372836 LOC105372836 -148.1kb Anti-sense Chr. 21 - 45055869 - 45073903 on NCBI Build GRCh38
          Downstream protein O-fucosyltransferase 2 POFUT2 C21orf80,
          FUT13
          37.4kb Anti-sense Chr. 21 - 45263928 - 45287912 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC2100007402.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Homo sapiens 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          adenosine to inosine editing
          mRNA processing
          negative regulation of cell population proliferation
          negative regulation of cell migration
          negative regulation of protein kinase activity by regulation of protein phosphorylation
          positive regulation of viral genome replication
          innate immune response
          defense response to virus
          regulation of cell cycle
          RNA binding
          double-stranded RNA adenosine deaminase activity
          mRNA binding
          protein binding
          metal ion binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Clariom D Assay PSR2100150132.hg.1 ENST00000348831.8 Ensembl
          Affymetrix Human Clariom D Assay PSR2100150132.hg.1 ENST00000360697.4 Ensembl
          Affymetrix Human Clariom D Assay PSR2100150132.hg.1 ENST00000389861.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2100150132.hg.1 ENST00000389863.8 Ensembl
          Affymetrix Human Clariom D Assay PSR2100150132.hg.1 ENST00000437626.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2100150132.hg.1 ENST00000492414.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2100150132.hg.1 ENST00000496664.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2100150132.hg.1 ENST00000629643.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2100150132.hg.1 NM_001112.3 Refseq
          Affymetrix Human Clariom D Assay PSR2100150132.hg.1 NM_001160230.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100150132.hg.1 NM_015833.3 Refseq
          Affymetrix Human Clariom D Assay PSR2100150132.hg.1 NM_015834.3 Refseq
          Affymetrix Human Clariom D Assay PSR2100150132.hg.1 NR_027672.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100150132.hg.1 NR_027673.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100150132.hg.1 NR_027674.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100150132.hg.1 NR_073200.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100150132.hg.1 TC2100007402.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2100150132.hg.1 ENST00000389861.7 Ensembl
          Affymetrix Human Clariom D Assay PSR2100150132.hg.1 ENST00000348831.8 Ensembl
          Affymetrix Human Clariom D Assay PSR2100150132.hg.1 ENST00000360697.4 Ensembl
          Affymetrix Human Clariom D Assay PSR2100150132.hg.1 ENST00000389863.8 Ensembl
          Affymetrix Human Clariom D Assay PSR2100150132.hg.1 ENST00000437626.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2100150132.hg.1 ENST00000492414.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2100150132.hg.1 ENST00000496664.5 Ensembl
          Affymetrix Human Clariom D Assay PSR2100150132.hg.1 ENST00000629643.2 Ensembl
          Affymetrix Human Clariom D Assay PSR2100150132.hg.1 NM_001112.3 Refseq
          Affymetrix Human Clariom D Assay PSR2100150132.hg.1 NM_001160230.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100150132.hg.1 NM_015833.3 Refseq
          Affymetrix Human Clariom D Assay PSR2100150132.hg.1 NM_015834.3 Refseq
          Affymetrix Human Clariom D Assay PSR2100150132.hg.1 NR_027672.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100150132.hg.1 NR_027673.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100150132.hg.1 NR_027674.1 Refseq
          Affymetrix Human Clariom D Assay PSR2100150132.hg.1 NR_073200.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Human Genome Survey Microarray Version 2.0 716112 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0