Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04333111_m1
          See other LOC100133050 GE Assays ›
          Gene Symbol:
          LOC100133050
          Gene Name:
          glucuronidase beta pseudogene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 100379505 - 100388254 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04333111_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC100133050
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027503.1 2 - 3 219 78
          GenBank mRNA BI830432.1 - 2 - 3 218 78
          BQ109458.1 - 2 - 3 219 78
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BF331307.1 - 1 - 2 Chr. 5 - 100066653 - 100067927 [+] on Build GRCh38 78

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream uncharacterized LOC102724855 CTD-2151A2.1 -488.7kb Sense Chr. 5 - 99533990 - 99577954 on NCBI Build GRCh38
          Downstream family with sequence similarity 174 member A FAM174A HGS_RE408,
          TMEM157,
          UNQ1912
          153.9kb Anti-sense Chr. 5 - 100535305 - 100586741 on NCBI Build GRCh38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0500011575.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC05001635.hg.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Homo sapiens Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 482368 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 944823 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 270410 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 268999 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 338200 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 56679 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P597190 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P39003 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 24761 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 17381 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 37915 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 19100 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_24_P597190 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_32_P39003 - -
          SurePrint G3 Human GE Microarray G4851A A_32_P39003 - -
          SurePrint G3 Human GE Microarray G4851A A_24_P597190 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04140322 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P04140322 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P04140321 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR05022250.hg.1 NR_027503.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05022250.hg.1 TC05001635.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR05022247.hg.1 NR_027503.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05022247.hg.1 TC05001635.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0500011573.hg.1 ENST00000621951.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011573.hg.1 ENST00000621951.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011573.hg.1 ENST00000621951.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011573.hg.1 ENST00000621951.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011573.hg.1 ENST00000621951.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR05022248.hg.1 NR_027503.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05022248.hg.1 TC05001635.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 PSR05022249.hg.1 NR_027503.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR05022249.hg.1 TC05001635.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC0500011575.hg.1 ENST00000621479.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011575.hg.1 ENST00000621479.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011575.hg.1 ENST00000621479.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011575.hg.1 ENST00000621479.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011575.hg.1 ENST00000621479.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011575.hg.1 ENST00000621479.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500011575.hg.1 NR_027503.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011575.hg.1 NR_027503.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011575.hg.1 NR_027503.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011575.hg.1 NR_027503.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011575.hg.1 NR_027503.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500011575.hg.1 NR_027503.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500008189.hg.1 ENST00000502652.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008189.hg.1 ENST00000502652.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008189.hg.1 ENST00000502652.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008189.hg.1 ENST00000502652.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008189.hg.1 ENST00000510302.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008189.hg.1 ENST00000510302.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0500008189.hg.1 NR_110562.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500008189.hg.1 NR_110562.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500008189.hg.1 NR_110562.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500008189.hg.1 NR_110562.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500008189.hg.1 NR_110562.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500008189.hg.1 NR_110562.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0500008189.hg.1 NR_110562.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011575.hg.1 ENST00000621479.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011575.hg.1 ENST00000621479.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011575.hg.1 ENST00000621479.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011575.hg.1 ENST00000621479.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011575.hg.1 ENST00000621479.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011575.hg.1 ENST00000621479.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011575.hg.1 NR_027503.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011575.hg.1 NR_027503.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011575.hg.1 NR_027503.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011575.hg.1 NR_027503.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011575.hg.1 NR_027503.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011575.hg.1 NR_027503.1 Refseq
          Affymetrix Human Clariom D Assay TC0500011573.hg.1 ENST00000621951.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011573.hg.1 ENST00000621951.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011573.hg.1 ENST00000621951.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011573.hg.1 ENST00000621951.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500011573.hg.1 ENST00000621951.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500008189.hg.1 ENST00000502652.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500008189.hg.1 ENST00000502652.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500008189.hg.1 ENST00000502652.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500008189.hg.1 ENST00000502652.2 Ensembl
          Affymetrix Human Clariom D Assay TC0500008189.hg.1 ENST00000510302.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500008189.hg.1 ENST00000510302.1 Ensembl
          Affymetrix Human Clariom D Assay TC0500008189.hg.1 NR_110562.1 Refseq
          Affymetrix Human Clariom D Assay TC0500008189.hg.1 NR_110562.1 Refseq
          Affymetrix Human Clariom D Assay TC0500008189.hg.1 NR_110562.1 Refseq
          Affymetrix Human Clariom D Assay TC0500008189.hg.1 NR_110562.1 Refseq
          Affymetrix Human Clariom D Assay TC0500008189.hg.1 NR_110562.1 Refseq
          Affymetrix Human Clariom D Assay TC0500008189.hg.1 NR_110562.1 Refseq
          Affymetrix Human Clariom D Assay TC0500008189.hg.1 NR_110562.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500180130.hg.1 NR_027503.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500180130.hg.1 TC0500011575.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500180130.hg.1 NR_027503.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500180133.hg.1 ENST00000621479.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500180133.hg.1 NR_027503.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500180133.hg.1 TC0500011575.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500180133.hg.1 ENST00000621479.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500180133.hg.1 NR_027503.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500180132.hg.1 ENST00000621479.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500180132.hg.1 NR_027503.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500180132.hg.1 TC0500011575.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500180132.hg.1 ENST00000621479.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500180132.hg.1 NR_027503.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500180129.hg.1 ENST00000621479.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500180129.hg.1 NR_027503.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500180129.hg.1 TC0500011575.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500180129.hg.1 ENST00000621479.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500180129.hg.1 NR_027503.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500180134.hg.1 NR_027503.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500180134.hg.1 TC0500011575.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500180134.hg.1 NR_027503.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500180131.hg.1 ENST00000621479.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500180131.hg.1 NR_027503.1 Refseq
          Affymetrix Human Clariom D Assay PSR0500180131.hg.1 TC0500011575.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0500180131.hg.1 ENST00000621479.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0500180131.hg.1 NR_027503.1 Refseq
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A027175 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A027175 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3239401 NR_027503.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3239401 BI830432.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3239401 BQ109458.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0