Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04333908_s1
          See other MIR34A GE Assays ›
          Gene Symbol:
          MIR34A MIR34AHG
          Gene Name:
          microRNA 34a
          MIR34A host gene
          Gene Aliases:
          MIRN34A, miRNA34A, mir-34, mir-34a
          -
          Chromosome Location:
          Chr.1: 9151668 - 9151777 on Build GRCh38
          Chromosome Location:
          Chr.1: 9148011 - 9182338 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs04333908_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MIR34A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_029610.1 1 64 104
          Gene Symbol: MIR34AHG
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_132742.1 2 454 104
          GenBank mRNA DB286351.1 - 2 455 104
          EF570048.1 - 2 221 104
          EF592573.1 - 2 509 104
          EF609116.1 - 2 454 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay is designed to target a precursor miRNA (pre-miRNA) target. It may not discriminate between the primary transcript (pri-miRNA) and the pre-miRNA for this miRNA. If you are looking for an assay to detect the mature miRNA, please search the MicroRNA collection of assays.
          This assay targets the same location as Clariom D transcript cluster(s) TC0100012791.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01002181.hg.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20046077 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20046050 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035925 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20046047 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20046046 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20046090 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20046029 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035938 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035900 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035918 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035936 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20046074 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20046027 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15021391 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035905 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035920 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035939 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20046081 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20046060 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20046080 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035941 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035916 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035914 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035899 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035908 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035940 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20046037 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035912 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035922 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20046057 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035909 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035931 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035937 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035924 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035901 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035913 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035896 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035921 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20046042 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20046091 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035900 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035939 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035918 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035912 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20046090 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035931 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035922 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20046074 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20046047 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035937 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20046091 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035896 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20046027 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20046060 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035901 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035941 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035913 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20046077 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20046037 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035924 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035908 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035914 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20046057 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035921 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20046042 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035905 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035916 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035936 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20046081 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035920 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20046029 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035938 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20046046 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20046050 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035909 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035899 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P15021391 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035940 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P20046080 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P00035925 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Human Gene 1.0 ST Array 7912249 - -
          Affymetrix Human Gene 1.0 ST Array 7912251 - -
          Affymetrix Human Gene 1.1 ST Array 7912248 - -
          Affymetrix Human Gene 1.1 ST Array 7912250 - -
          Affymetrix Human Transcriptome Array 2.0 TC0100012791.hg.1 ENST00000385130.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012791.hg.1 ENST00000437157.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012791.hg.1 ENST00000437157.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012791.hg.1 ENST00000437157.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012791.hg.1 ENST00000437157.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012791.hg.1 ENST00000437157.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100012791.hg.1 NR_029610.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01033617.hg.1 ENST00000385130.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01033617.hg.1 NR_029610.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01033617.hg.1 TC01002181.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0100012791.hg.1 ENST00000385130.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100012791.hg.1 ENST00000437157.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100012791.hg.1 ENST00000437157.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100012791.hg.1 ENST00000437157.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100012791.hg.1 ENST00000437157.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100012791.hg.1 ENST00000437157.2 Ensembl
          Affymetrix Human Clariom D Assay TC0100012791.hg.1 NR_029610.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100194607.hg.1 ENST00000385130.1 Ensembl
          Affymetrix Human Clariom D Assay PSR0100194607.hg.1 NR_029610.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100194607.hg.1 TC0100012791.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100194607.hg.1 NR_029610.1 Refseq
          Affymetrix Human Clariom D Assay PSR0100194607.hg.1 ENST00000385130.1 Ensembl
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_3308455 NR_029610.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3308455 EF592573.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_3308455 EF609116.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3308455 NR_029610.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3308455 EF592573.1 human_transcriptome
          HumanHT-12 V4.0 expression beadchip ILMN_3308455 EF609116.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3308455 NR_029610.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3308455 EF592573.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 expression beadchip ILMN_3308455 EF609116.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3308455 NR_029610.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3308455 EF592573.1 human_transcriptome
          Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip ILMN_3308455 EF609116.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0