Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000083.hg.1 |
ENST00000215530.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000083.hg.1 |
NM_020637.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000083.hg.1 |
TC19000011.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006475.hg.1 |
ENST00000215530.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006475.hg.1 |
ENST00000215530.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006475.hg.1 |
ENST00000215530.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006475.hg.1 |
ENST00000215530.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006475.hg.1 |
ENST00000215530.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006475.hg.1 |
ENST00000215530.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006475.hg.1 |
ENST00000586042.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006475.hg.1 |
ENST00000586042.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006475.hg.1 |
ENST00000586042.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006475.hg.1 |
ENST00000586042.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006475.hg.1 |
ENST00000586042.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006475.hg.1 |
ENST00000586042.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006475.hg.1 |
ENST00000591390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006475.hg.1 |
ENST00000591390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006475.hg.1 |
ENST00000591390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006475.hg.1 |
NM_001300812.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006475.hg.1 |
NM_001300812.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006475.hg.1 |
NM_001300812.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006475.hg.1 |
NM_001300812.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006475.hg.1 |
NM_001300812.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006475.hg.1 |
NM_020637.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006475.hg.1 |
NM_020637.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006475.hg.1 |
NM_020637.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006475.hg.1 |
ENST00000215530.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006475.hg.1 |
ENST00000215530.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006475.hg.1 |
ENST00000215530.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006475.hg.1 |
ENST00000215530.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006475.hg.1 |
ENST00000215530.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006475.hg.1 |
ENST00000215530.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006475.hg.1 |
ENST00000586042.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006475.hg.1 |
ENST00000586042.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006475.hg.1 |
ENST00000586042.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006475.hg.1 |
ENST00000586042.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006475.hg.1 |
ENST00000586042.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006475.hg.1 |
ENST00000586042.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006475.hg.1 |
ENST00000591390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006475.hg.1 |
ENST00000591390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006475.hg.1 |
ENST00000591390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006475.hg.1 |
NM_001300812.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006475.hg.1 |
NM_001300812.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006475.hg.1 |
NM_001300812.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006475.hg.1 |
NM_001300812.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006475.hg.1 |
NM_001300812.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006475.hg.1 |
NM_020637.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006475.hg.1 |
NM_020637.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006475.hg.1 |
NM_020637.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144336.hg.1 |
ENST00000215530.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144336.hg.1 |
ENST00000586042.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144336.hg.1 |
ENST00000591390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144336.hg.1 |
NM_001300812.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144336.hg.1 |
NM_020637.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144336.hg.1 |
TC1900006475.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900144336.hg.1 |
NM_001300812.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144336.hg.1 |
ENST00000215530.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144336.hg.1 |
ENST00000586042.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144336.hg.1 |
ENST00000591390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144336.hg.1 |
NM_020637.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144335.hg.1 |
ENST00000215530.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144335.hg.1 |
ENST00000586042.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144335.hg.1 |
ENST00000591390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144335.hg.1 |
NM_001300812.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900144335.hg.1 |
TC1900006475.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900144335.hg.1 |
ENST00000215530.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144335.hg.1 |
ENST00000586042.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144335.hg.1 |
ENST00000591390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900144335.hg.1 |
NM_001300812.1 |
Refseq |