Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04398527_m1
          See other TPM1 GE Assays ›
          Gene Symbol:
          TPM1
          Gene Name:
          tropomyosin 1 (alpha)
          Gene Aliases:
          C15orf13, CMD1Y, CMH3, HEL-S-265, HTM-alpha, LVNC9, TMSA
          Chromosome Location:
          Chr.15: 63042196 - 63071915 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04398527_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TPM1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001018004.1 NP_001018004.1 6 - 7 837 99
          NM_001018005.1 NP_001018005.1 6 - 7 837 99
          NM_001018007.1 NP_001018007.1 6 - 7 837 99
          NM_001018008.1 NP_001018008.1 5 - 6 677 99
          NM_001301244.1 NP_001288173.1 6 - 7 837 99
          NM_001301289.1 NP_001288218.1 5 - 6 677 99
          XM_005254637.1 XP_005254694.1 7 - 8 918 99
          XM_005254638.3 XP_005254695.2 6 - 7 1280 99
          XM_005254645.2 XP_005254702.2 6 - 7 1280 99
          XM_005254647.3 XP_005254704.1 5 - 6 663 99
          XM_017022534.1 XP_016878023.1 6 - 7 1280 99
          XM_017022535.1 XP_016878024.1 6 - 7 792 99
          XM_017022540.1 XP_016878029.1 5 - 6 663 99
          XM_017022541.1 XP_016878030.1 5 - 6 663 99
          GenBank mRNA AB209041.1 - 5 - 6 714 99
          AK057989.1 - 8 - 9 623 99
          AK129882.1 - 5 - 6 524 99
          AK131384.1 - 7 - 8 851 99
          AK299387.1 - 6 - 7 726 99
          AK301319.1 - 5 - 6 573 99
          AL050179.1 - 6 - 7 718 99
          BC050473.1 - 5 - 6 664 99
          BC053545.1 - 5 - 6 675 5730451 99
          CD679271.1 - 2 - 3 168 99
          KU178367.1 - 6 - 7 646 99
          M19713.1 - 6 - 7 702 99
          M19714.1 - 6 - 7 588 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC1500007510.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC15000578.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TPM1

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          positive regulation of heart rate by epinephrine
          movement of cell or subcellular component
          muscle contraction
          regulation of muscle contraction
          cytoskeleton organization
          regulation of heart contraction
          regulation of cell shape
          muscle filament sliding
          negative regulation of cell migration
          ruffle organization
          positive regulation of ATPase activity
          cellular response to reactive oxygen species
          wound healing
          sarcomere organization
          positive regulation of cell adhesion
          positive regulation of stress fiber assembly
          ventricular cardiac muscle tissue morphogenesis
          cardiac muscle contraction
          negative regulation of vascular associated smooth muscle cell proliferation
          negative regulation of vascular associated smooth muscle cell migration
          actin binding
          structural constituent of cytoskeleton
          protein binding
          cytoskeletal protein binding
          structural constituent of muscle

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 PSR15004610.hg.1 ENST00000267996.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15004610.hg.1 ENST00000288398.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15004610.hg.1 ENST00000317516.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15004610.hg.1 ENST00000334895.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15004610.hg.1 ENST00000357980.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15004610.hg.1 ENST00000358278.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15004610.hg.1 ENST00000403994.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15004610.hg.1 ENST00000404484.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15004610.hg.1 ENST00000559281.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15004610.hg.1 ENST00000559397.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15004610.hg.1 ENST00000559556.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15004610.hg.1 ENST00000560959.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR15004610.hg.1 NM_000366.5 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15004610.hg.1 NM_001018004.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15004610.hg.1 NM_001018005.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15004610.hg.1 NM_001018006.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15004610.hg.1 NM_001018007.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15004610.hg.1 NM_001018008.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15004610.hg.1 NM_001018020.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR15004610.hg.1 TC15000578.hg.1 HTA TC
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000267996.11 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000288398.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000317516.11 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000334895.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000357980.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000358278.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000403994.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000404484.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000558072.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000558264.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000558347.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000558544.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000559108.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000559281.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000559397.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000559556.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000559831.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000560615.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000560959.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000560970.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000560975.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000561242.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000561266.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000561395.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 NM_000366.5 Refseq
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 NM_001018004.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 NM_001018005.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 NM_001018006.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 NM_001018007.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 NM_001018008.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 NM_001018020.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 NM_001301244.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 NM_001301289.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 TC1500007510.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000334895.9 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000267996.11 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000288398.10 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000317516.11 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000357980.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000358278.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000403994.7 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000404484.8 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000558072.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000558264.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000558347.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000558544.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000559108.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000559281.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000559397.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000559556.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000559831.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000560615.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000560959.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000560970.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000560975.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000561242.1 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000561266.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 ENST00000561395.5 Ensembl
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 NM_000366.5 Refseq
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 NM_001018004.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 NM_001018005.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 NM_001018006.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 NM_001018007.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 NM_001018008.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 NM_001018020.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 NM_001301244.1 Refseq
          Affymetrix Human Clariom D Assay PSR1500153174.hg.1 NM_001301289.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0