Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs04399143_g1
See other PRPF40B GE Assays ›
Gene Symbol:
PRPF40B
Gene Name:
pre-mRNA processing factor 40 homolog B
Gene Aliases:
HYPC
Chromosome Location:
Chr.12: 49622717 - 49644669 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs04399143_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: PRPF40B
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001031698.2
NP_001026868.2
12 - 13
1178
78
NM_012272.2
NP_036404.1
11 - 12
1468
78
XM_006719324.3
XP_006719387.1
11 - 12
2232
78
XM_006719325.3
XP_006719388.1
11 - 12
2232
78
XM_011538135.2
XP_011536437.1
11 - 12
2232
78
XM_011538136.2
XP_011536438.1
11 - 12
2232
78
XM_011538137.2
XP_011536439.1
10 - 11
2151
78
XM_011538138.2
XP_011536440.1
10 - 11
2121
78
XM_011538139.2
XP_011536441.1
12 - 13
1172
78
XM_011538140.2
XP_011536442.1
12 - 13
1172
78
XM_011538141.1
XP_011536443.1
11 - 12
998
78
XM_011538143.2
XP_011536445.1
11 - 12
1014
78
XM_011538144.2
XP_011536446.1
10 - 11
887
78
XM_017019135.1
XP_016874624.1
11 - 12
935
78
XM_017019136.1
XP_016874625.1
10 - 11
887
78
XM_017019137.1
XP_016874626.1
10 - 11
887
78
XM_017019138.1
XP_016874627.1
11 - 12
2232
78
XM_017019139.1
XP_016874628.1
7 - 8
898
78
GenBank mRNA
AL834216.1
-
11 - 12
1468
78
BC031917.1
-
9 - 10
774
78
BC050398.2
-
12 - 13
1014
78
BC067364.1
-
11 - 12
999
6644672
78
Back To Top
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017019139.1 transcripts.
Set Membership:
Ambion siRNA available
Amplicon length between 71 and 85
Mammalian Gene Collection
Detect Genomic DNA
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene PRPF40B
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
RNA binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0