Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs04399403_m1
See other ARNTL GE Assays ›
Gene Symbol:
ARNTL
Gene Name:
aryl hydrocarbon receptor nuclear translocator like
Gene Aliases:
BMAL1, BMAL1c, JAP3, MOP3, PASD3, TIC, bHLHe5
Chromosome Location:
Chr.11: 13276613 - 13387268 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs04399403_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_Primer Limited | S: 360 rxns
FAM-MGB_Primer Limited | S: 360 rxns
ABY-MGB_Primer Limited | S: 360 rxns
JUN-MGB_Primer Limited | S: 360 rxns
CY5-MGB_Primer Limited | S: 360 rxns
VIC-MGB_Primer Limited | M: 750 rxns
FAM-MGB_Primer Limited | M: 750 rxns
ABY-MGB_Primer Limited | M: 750 rxns
JUN-MGB_Primer Limited | M: 750 rxns
CY5-MGB_Primer Limited | M: 750 rxns
VIC-MGB_Primer Limited | L: 2900 rxns
FAM-MGB_Primer Limited | L: 2900 rxns
ABY-MGB_Primer Limited | L: 2900 rxns
JUN-MGB_Primer Limited | L: 2900 rxns
CY5-MGB_Primer Limited | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ARNTL
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001030272.2
NP_001025443.1
7 - 8
790
82
NM_001030273.2
NP_001025444.1
8 - 9
861
82
NM_001178.5
NP_001169.3
8 - 9
845
82
NM_001297719.1
NP_001284648.1
8 - 9
845
82
NM_001297722.1
NP_001284651.1
7 - 8
790
82
NM_001297724.1
NP_001284653.1
8 - 9
854
82
XM_006718233.3
XP_006718296.1
7 - 8
732
82
XM_006718234.1
XP_006718297.1
8 - 9
798
82
XM_011520105.2
XP_011518407.2
6 - 7
896
82
XM_011520107.2
XP_011518409.2
6 - 7
891
82
XM_011520108.1
XP_011518410.1
8 - 9
838
82
XM_011520109.1
XP_011518411.1
6 - 7
593
82
XM_011520110.1
XP_011518412.1
9 - 10
853
82
XM_011520111.1
XP_011518413.1
7 - 8
668
82
XM_011520112.2
XP_011518414.1
9 - 10
853
82
XM_011520113.1
XP_011518415.1
9 - 10
853
82
XM_017017738.1
XP_016873227.1
6 - 7
894
82
XM_017017739.1
XP_016873228.1
6 - 7
890
82
XM_017017740.1
XP_016873229.1
7 - 8
732
82
XM_017017741.1
XP_016873230.1
6 - 7
593
82
XM_017017742.1
XP_016873231.1
6 - 7
593
82
XM_017017743.1
XP_016873232.1
6 - 7
593
82
XM_017017744.1
XP_016873233.1
9 - 10
853
82
XM_017017745.1
XP_016873234.1
8 - 9
798
82
XM_017017746.1
XP_016873235.1
8 - 9
808
82
XM_017017747.1
XP_016873236.1
7 - 8
668
82
XM_017017748.1
XP_016873237.1
7 - 8
725
82
GenBank mRNA
AB000812.1
-
5 - 6
426
82
AB000815.1
-
6 - 7
497
82
AB000816.1
-
7 - 8
837
82
AF044288.1
-
8 - 9
755
82
AK095749.1
-
8 - 9
746
82
AK291510.1
-
7 - 8
703
82
AK315174.1
-
8 - 9
783
82
BC016674.1
-
5 - 6
389
3921870
82
BC031214.1
-
8 - 9
686
4793007
82
BC041129.1
-
8 - 9
769
5753529
82
D89722.1
-
5 - 6
402
82
U51627.1
-
2 - 3
384
82
U60415.1
-
7 - 8
689
82
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length between 71 and 85
Mammalian Gene Collection
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene ARNTL
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
transcription by RNA polymerase II
spermatogenesis
circadian rhythm
negative regulation of TOR signaling
circadian regulation of gene expression
regulation of protein catabolic process
regulation of hair cycle
positive regulation of circadian rhythm
proteasome-mediated ubiquitin-dependent protein catabolic process
negative regulation of fat cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription by RNA polymerase II
regulation of neurogenesis
regulation of insulin secretion
regulation of cell cycle
response to redox state
maternal process involved in parturition
positive regulation of canonical Wnt signaling pathway
oxidative stress-induced premature senescence
regulation of type B pancreatic cell development
negative regulation of glucocorticoid receptor signaling pathway
regulation of cellular senescence
positive regulation of skeletal muscle cell differentiation
transcription cis-regulatory region binding
core promoter sequence-specific DNA binding
DNA binding
protein binding
aryl hydrocarbon receptor binding
bHLH transcription factor binding
sequence-specific DNA binding
protein heterodimerization activity
Hsp90 protein binding
E-box binding
DNA-binding transcription factor binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0