Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04402112_g1
          See other CCDC115 GE Assays ›
          Gene Symbol:
          CCDC115
          Gene Name:
          coiled-coil domain containing 115
          Gene Aliases:
          CDG2O, ccp1
          Chromosome Location:
          Chr.2: 130337933 - 130342681 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04402112_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CCDC115
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001321118.1 NP_001308047.1 1 - 2 545 102
          NR_104471.1 1 - 2 545 102
          GenBank mRNA AK295922.1 - 1 - 2 202 102
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AL557449.3 - 1 - 2 188 Chr. 2 - 130338827 - 130342324 [-] on Build GRCh38 102
          GenBankmRNA HY170661.1 - 1 - 2 114 Chr. 2 - 130339155 - 130342250 [-] on Build GRCh38 102
          GenBankmRNA BI553860.1 - 191 Chr. - - [-] on Build 102
          GenBankmRNA DR000373.1 - 1 - 2 174 Chr. 2 - 130339021 - 130342310 [-] on Build GRCh38 102
          GenBankmRNA BI915687.1 - 1 - 2 467 Chr. 2 - 130340947 - 130342603 [-] on Build GRCh38 102
          GenBankmRNA DC338850.1 - 1 - 2 138 Chr. 2 - 130339679 - 130342274 [-] on Build GRCh38 102
          GenBankmRNA BI919206.1 - 1 - 2 493 Chr. 2 - 130339183 - 130342628 [-] on Build GRCh38 102
          GenBankmRNA DC336736.1 - 1 - 2 202 Chr. 2 - 130339705 - 130342338 [-] on Build GRCh38 102
          GenBankmRNA DC337825.1 - 1 - 2 514 Chr. 2 - 130341907 - 130342650 [-] on Build GRCh38 102
          GenBankmRNA DC305982.1 - 1 - 2 185 Chr. 2 - 130340940 - 130342321 [-] on Build GRCh38 102
          GenBankmRNA DC337793.1 - 1 - 2 514 Chr. 2 - 130341871 - 130342650 [-] on Build GRCh38 102
          GenBankmRNA HY004061.1 - 1 - 2 119 Chr. 2 - 130339176 - 130342255 [-] on Build GRCh38 102
          GenBankmRNA BI910277.1 - 1 - 2 197 Chr. 2 - 130339704 - 130342329 [-] on Build GRCh38 102

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream NOC2 like nucleolar associated transcriptional repressor pseudogene 1 NOC2LP1 -109.8kb Anti-sense Chr. 2 - 130229320 - 130232086 on NCBI Build GRCh38
          Downstream IMP4 homolog,
          U3 small nucleolar ribonucleoprotein
          IMP4 BXDC4 216.0 Anti-sense Chr. 2 - 130342225 - 130347961 on NCBI Build GRCh38

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0