Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012221.hg.1 |
ENST00000515306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012221.hg.1 |
ENST00000515306.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012221.hg.1 |
NR_105052.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012221.hg.1 |
NR_105052.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012221.hg.1 |
NR_105052.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012221.hg.1 |
NR_105053.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012221.hg.1 |
NR_105053.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500184831.hg.1 |
ENST00000515306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184831.hg.1 |
NR_105052.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500184831.hg.1 |
NR_105053.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500184831.hg.1 |
TC0500012221.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500184831.hg.1 |
NR_105053.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500184831.hg.1 |
ENST00000515306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500184831.hg.1 |
NR_105052.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012221.hg.1 |
ENST00000515306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012221.hg.1 |
ENST00000515306.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012221.hg.1 |
NR_105052.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012221.hg.1 |
NR_105052.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012221.hg.1 |
NR_105052.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012221.hg.1 |
NR_105053.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012221.hg.1 |
NR_105053.2 |
Refseq |