Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR6072 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_106720.1 | 1 | 37 | 56 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 700 | LINC00700 | -61.7kb | Sense | Chr. 10 - 2005473 - 2014348 on NCBI Build GRCh38 | |
Downstream | long intergenic non-protein coding RNA 701 | LINC00701 | 224.2kb | Sense | Chr. 10 - 2300319 - 2315074 on NCBI Build GRCh38 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |