Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012387.hg.1 |
ENST00000300167.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012387.hg.1 |
ENST00000300167.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012387.hg.1 |
ENST00000300167.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012387.hg.1 |
ENST00000300167.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012387.hg.1 |
ENST00000362219.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012387.hg.1 |
ENST00000362219.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012387.hg.1 |
ENST00000553459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012387.hg.1 |
ENST00000553459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012387.hg.1 |
ENST00000553459.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012387.hg.1 |
NR_030170.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012387.hg.1 |
NR_030170.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012387.hg.1 |
NR_108078.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012387.hg.1 |
NR_108078.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012387.hg.1 |
NR_108078.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012387.hg.1 |
NR_108078.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012387.hg.1 |
NR_108078.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012387.hg.1 |
NR_108079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012387.hg.1 |
NR_108079.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012387.hg.1 |
NR_108079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000184491.hg.1 |
ENST00000300167.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000184491.hg.1 |
NR_108078.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000184491.hg.1 |
TC1000012387.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000184491.hg.1 |
NR_108078.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000184491.hg.1 |
ENST00000300167.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000184492.hg.1 |
ENST00000300167.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000184492.hg.1 |
ENST00000553459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000184492.hg.1 |
NR_108078.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000184492.hg.1 |
NR_108079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000184492.hg.1 |
TC1000012387.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000184492.hg.1 |
ENST00000300167.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000184492.hg.1 |
ENST00000553459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000184492.hg.1 |
NR_108078.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000184492.hg.1 |
NR_108079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012387.hg.1 |
ENST00000300167.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012387.hg.1 |
ENST00000300167.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012387.hg.1 |
ENST00000300167.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012387.hg.1 |
ENST00000300167.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012387.hg.1 |
ENST00000362219.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012387.hg.1 |
ENST00000362219.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012387.hg.1 |
ENST00000553459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012387.hg.1 |
ENST00000553459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012387.hg.1 |
ENST00000553459.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012387.hg.1 |
NR_030170.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012387.hg.1 |
NR_030170.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012387.hg.1 |
NR_108078.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012387.hg.1 |
NR_108078.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012387.hg.1 |
NR_108078.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012387.hg.1 |
NR_108078.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012387.hg.1 |
NR_108078.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012387.hg.1 |
NR_108079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012387.hg.1 |
NR_108079.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012387.hg.1 |
NR_108079.1 |
Refseq |