Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010000.hg.1 |
ENST00000419662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010000.hg.1 |
ENST00000419662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010000.hg.1 |
ENST00000419662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010000.hg.1 |
ENST00000419662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010000.hg.1 |
ENST00000419662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010000.hg.1 |
ENST00000419662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010000.hg.1 |
ENST00000419662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010000.hg.1 |
ENST00000419662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010000.hg.1 |
ENST00000440595.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010000.hg.1 |
ENST00000440595.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010000.hg.1 |
ENST00000440595.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010000.hg.1 |
ENST00000440595.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010000.hg.1 |
ENST00000440595.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010000.hg.1 |
ENST00000440595.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010000.hg.1 |
ENST00000440595.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010000.hg.1 |
ENST00000440595.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010000.hg.1 |
ENST00000440595.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010000.hg.1 |
ENST00000440595.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010000.hg.1 |
NR_109885.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010000.hg.1 |
NR_109885.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010000.hg.1 |
NR_109885.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010000.hg.1 |
NR_109885.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010000.hg.1 |
NR_109885.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010000.hg.1 |
NR_109886.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010000.hg.1 |
NR_109886.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010000.hg.1 |
NR_109886.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000010000.hg.1 |
NR_109886.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000171361.hg.1 |
ENST00000419662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171361.hg.1 |
ENST00000419662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171361.hg.1 |
ENST00000440595.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171361.hg.1 |
ENST00000440595.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171361.hg.1 |
NR_109885.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000171361.hg.1 |
NR_109886.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000171361.hg.1 |
TC2000010000.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000171361.hg.1 |
ENST00000440595.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171361.hg.1 |
ENST00000419662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171361.hg.1 |
ENST00000419662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171361.hg.1 |
ENST00000440595.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171361.hg.1 |
NR_109885.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000171361.hg.1 |
NR_109886.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000010000.hg.1 |
ENST00000419662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010000.hg.1 |
ENST00000419662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010000.hg.1 |
ENST00000419662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010000.hg.1 |
ENST00000419662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010000.hg.1 |
ENST00000419662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010000.hg.1 |
ENST00000419662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010000.hg.1 |
ENST00000419662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010000.hg.1 |
ENST00000419662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010000.hg.1 |
ENST00000440595.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010000.hg.1 |
ENST00000440595.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010000.hg.1 |
ENST00000440595.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010000.hg.1 |
ENST00000440595.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010000.hg.1 |
ENST00000440595.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010000.hg.1 |
ENST00000440595.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010000.hg.1 |
ENST00000440595.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010000.hg.1 |
ENST00000440595.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010000.hg.1 |
ENST00000440595.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010000.hg.1 |
ENST00000440595.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000010000.hg.1 |
NR_109885.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000010000.hg.1 |
NR_109885.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000010000.hg.1 |
NR_109885.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000010000.hg.1 |
NR_109885.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000010000.hg.1 |
NR_109885.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000010000.hg.1 |
NR_109886.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000010000.hg.1 |
NR_109886.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000010000.hg.1 |
NR_109886.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000010000.hg.1 |
NR_109886.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000171360.hg.1 |
ENST00000419662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171360.hg.1 |
ENST00000419662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171360.hg.1 |
ENST00000440595.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171360.hg.1 |
ENST00000440595.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171360.hg.1 |
NR_109885.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000171360.hg.1 |
NR_109886.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000171360.hg.1 |
TC2000010000.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000171360.hg.1 |
ENST00000419662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171360.hg.1 |
ENST00000419662.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171360.hg.1 |
ENST00000440595.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171360.hg.1 |
ENST00000440595.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000171360.hg.1 |
NR_109885.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000171360.hg.1 |
NR_109886.1 |
Refseq |